Skip to Content
Merck
All Photos(5)

Documents

WH0006421M2

Sigma-Aldrich

Monoclonal Anti-SFPQ antibody produced in mouse

clone 6D7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-POMP100, Anti-PSF, Anti-splicing factor proline/glutamine-rich (polypyrimidine tract binding protein associated)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6D7, monoclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SFPQ(6421)

General description

Splicing factor proline and glutamine rich (SFPQ), also known as polypyrimidine tract-binding protein-associated-splicing factor (PSF), is a multifunctional nuclear protein. The protein is characterized with an N-terminal glycine rich domain, a proline/glutamine-rich domain (P/Q), two RNA recognition motifs (RRMs) and a C-terminal region with two nuclear localization signals. The SFPQ gene is mapped to human chromosome 1p34.

Immunogen

SFPQ (NP_005057, 269 a.a. ~ 361 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
EEKISDSEGFKANLSLLRRPGEKTYTQRCRLFVGNLPADITEDEFKRLFAKYGEPGEVFINKGKGFGFIKLESRALAEIAKAELDDTPMRGRQ

Application

Monoclonal Anti-SFPQ antibody produced in mouse has been used in Western Blotting and immunofluorescence.

Biochem/physiol Actions

Splicing factor proline and glutamine rich (SFPQ), along with its binding partner non-POU domain-containing octamer-binding protein (NONO/p54nrb), plays a vital role in RNA processing, RNA splicing and transcriptional regulation. Additionally, these proteins also play a regulatory role in selective nuclear retention of defective mRNAs. SFPQ participates in transcription repression by recruiting transcription regulator proteins Sin3a and histone deacetylase (HDAC).

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arginine methylation and citrullination of splicing factor proline- and glutamine-rich (SFPQ/PSF) regulates its association with mRNA
Ambrosius P. Snijders
RNA (2015)
The t(1;9)(p34;q34) and t(8;12)(p11;q15) fuse pre-mRNA processing proteins SFPQ (PSF) and CPSF6 to ABL and FGFR1
Claire H
Genes Chromosomes Cancer (2008)
Cell-type specific role of the RNA-binding protein, NONO, in the DNA double-strand break response in the mouse testes
DNA Repair (2017)
Paclitaxel Reduces Axonal Bclw to Initiate IP3R1-Dependent Axon Degeneration
Sarah E.Pease-Raissi
Neuron (2017)
Stefan G Kathman et al.
Nature chemical biology, 19(7), 825-836 (2023-03-03)
Much of the human proteome is involved in mRNA homeostasis, but most RNA-binding proteins lack chemical probes. Here we identify electrophilic small molecules that rapidly and stereoselectively decrease the expression of transcripts encoding the androgen receptor and its splice variants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service