Skip to Content
Merck
All Photos(2)

Key Documents

WG16

Sigma-Aldrich

Wright-Giemsa Stain, Modified

Wright-Giemsa Stain, Modified
Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116121
eCl@ss:
42039003
NACRES:
NA.47
Pricing and availability is not currently available.

form

solution

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

0.4 % (w/v) in methanol (buffered at pH 6.8)

pH

6.7-6.9

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

When blood films are stained using Wright-Giemsa Stain, the white blood cell nucleus and cytoplasm take on the characteristic blue or pink coloration. The combination of purified eosin and thiazine dyes in the product eliminates inconsistent staining and yields reproducible chromogenic responses.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shuyuan Liu et al.
International journal of medical sciences, 15(7), 674-681 (2018-06-19)
The cullin-RING ligase (CRL)-NEDD8 pathway maintains essential cellular processes, including cell cycle progression, apoptosis, autophagy, DNA repair, antigen processing and signal transduction. Growing evidence demonstrates that the alteration of the CRL-NEDD8 pathway in some cancers constitutes an attractive target for
Limor Shtoots et al.
Neurobiology of learning and memory, 154, 12-21 (2017-10-01)
Previous studies indicate that individuals exposed to stress in juvenility are more prone to suffer from stress-related psychopathologies in adulthood. Evidence suggests that exposure to enriched environment (EE) conditions alleviates juvenile stress (JVS) effects. Exposure to stress has been found
Zhenfan Yang et al.
Science translational medicine, 8(368), 368ra172-368ra172 (2016-12-09)
Non-small-cell lung cancer patients with activating mutations in epidermal growth factor receptor (EGFR) respond to EGFR tyrosine kinase inhibitor (TKI) treatment. Nevertheless, patients often develop central nervous system (CNS) metastases during treatment, even when their extracranial tumors are still under
Mary P Lee et al.
Blood, 130(23), 2537-2547 (2017-10-12)
The orphan nuclear receptors TR4 (NR2C2) and TR2 (NR2C1) are the DNA-binding subunits of the macromolecular complex, direct repeat erythroid-definitive, which has been shown to repress
Inhibition of CRL-NEDD8 pathway as a new approach to enhance ATRA-induced differentiation of acute promyelocytic leukemia cells
Liu S, et al.
International Journal of Medical Sciences, 15(7), 674-674 (2018)

Questions

  1. What is the preservation process for slides stained with the Wright-Giemsa stain if they cannot be viewed immediately?

    1 answer
    1. the Wright Giemsa Modified solution is intended for staining peripheral blood or bone marrow smears, without mention of staining tissue sections. If staining smears, simply complete the staining procedure as directed in the IFU, then store the dried slides in a slide folder or box without coverslipping, as coverslipping is not recommended and is not a common practice in hospital settings.

      For long-term storage, slides are typically stored in slide file cabinets, and oil immersion is usually not required. However, if slides need to be viewed under oil immersion, a coverslip can be placed atop the stained slide, and once viewed, the coverslip can be removed and discarded. Any excess oil can be absorbed using a Kimwipe or tissue paper without wiping, as wiping may remove stained cells from the slide.

      Although no formal studies have been conducted to determine the longevity of stained slides, it is expected that the staining should be adequate for several weeks or months. For longer retention, it is suggested to take pictures of several fields from the slides for archival purposes.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service