Skip to Content
Merck

Skip To

V9380

Freezing vials 2.0 mL, sterile

natural cap

Synonym(s):

cryopreservation tubes, cryopreservation vials, cryovials, freezing tubes, Simport Plastics T334-6SPRNSI

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NB.22
UNSPSC Code:
41121704
Capacity:
2 mL
Material:
polypropylene
Sterility:
sterile; γ-irradiated (Certified RNase-, DNase-, Pyrogen- and DNA-free.), sterile; γ-irradiated
Packaging:
pack of 100 ea
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polypropylene

sterility

sterile; γ-irradiated (Certified RNase-, DNase-, Pyrogen- and DNA-free.), sterile; γ-irradiated

packaging

pack of 100 ea

manufacturer/tradename

Simport T334-6SPRNSI

technique(s)

cryopreservation: suitable

capacity

2 mL

color

natural cap

General description

Cryogenic vials, 2.0 mL are designed for storing biological material, human or animal cells, at temperatures as low as -196 °C. In ["Micrewtube"] with washer seal screw cap, the washer seal is secured in the top of the cap and this ensures a positive leakproof seal, time after time, thereby keeping the integrity of the small sample under even the most adverse conditions.

Application

Freezing vials 2.0 mL, sterile has been used to store cerebrospinal fluid (CSF). It has also been used for agarose embedment and sectioning of rice roots harvested from petri-plate assay.

Features and Benefits

  • Graduated with marking square for sample identification
  • Color coded caps have internal threads and O-ring seal to minimize contamination
  • Do not immerse in the liquid phase of liquid nitrogen
  • Made of polypropylene
  • Autoclavable
  • Perfect for cryogenic work


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
V9255V5005V5381
material

polypropylene

material

polypropylene

material

polypropylene

material

colorless tube, polypropylene tube, skirted bottom

manufacturer/tradename

Simport T334-6SPRNSI

manufacturer/tradename

Simport T334-6SPRBSI

manufacturer/tradename

Simport T334-6SPRYSI

manufacturer/tradename

-

sterility

sterile; γ-irradiated (Certified RNase-, DNase-, Pyrogen- and DNA-free.)

sterility

sterile; γ-irradiated (Certified RNase-, DNase-, Pyrogen- and DNA-free.)

sterility

sterile; γ-irradiated (Certified RNase-, DNase-, Pyrogen- and DNA-free.)

sterility

sterile

capacity

2 mL

capacity

2 mL

capacity

2 mL

capacity

2 mL

packaging

pack of 100 ea

packaging

pack of 100 ea

packaging

pack of 100 ea

packaging

pack of 500 ea (10 x 50 ea)

color

natural cap

color

blue cap

color

yellow cap

color

natural Cap


Still not finding the right product?

Explore all of our products under Freezing vials 2.0 mL, sterile




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library