V5005
Freezing vials 2.0 mL, sterile
yellow cap
Synonym(s):
cryopreservation tubes, cryopreservation vials, cryovials, freezing tubes
About This Item
Recommended Products
sterility
sterile; γ-irradiated (Certified RNase-, DNase-, Pyrogen- and DNA-free.)
packaging
pack of 100 ea
manufacturer/tradename
Simport T334-6SPRYSI
technique(s)
cryopreservation: suitable
capacity
2 mL
color
yellow cap
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Graduated with marking square for sample identification.
- Color coded caps have internal threads and O-ring seal to minimize contamination.
- Do not immerse in the liquid phase of liquid nitrogen.
- Made of polypropyleneAutoclavablePerfect for cryogenic work
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service