Skip to Content
Merck
All Photos(5)

Documents

V9264

Sigma-Aldrich

Anti-Vinculin antibody, Mouse monoclonal

clone hVIN-1, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-Vinculin antibody produced in mouse

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

hVIN-1, monoclonal

form

buffered aqueous solution

mol wt

antigen 116 kDa

species reactivity

frog, chicken, mouse, canine, human, bovine, rat, turkey

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunocytochemistry: 5-10 μg/mL using HS-68 human fibroblast cell culture
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: 0.05-0.1 μg/mL using HS-68 total cell extract

isotype

IgG1

UniProt accession no.

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VCL(7414)
mouse ... Vcl(22330)
rat ... Vcl(305679)

Looking for similar products? Visit Product Comparison Guide

General description

Anti-Vinculin antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the hVIN-1 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with vinculin purified from human uterus. Vinculin is a cytoskeletal protein associated with the cytoplasmic faces of both cell-cell and cell-extracellular matrix adherens-type junctions. In muscle, vinculin is localized in the fascia adherens of the intercalated disk (cardiac muscle), myotendinous junctions (skeletal muscle), neuromuscular junctions, and the membrane associated dense bodies of smooth muscle.

Specificity

Specifically labels vinculin at cell-cell and cell-substrate contacts. Reacts strongly with human vinculin. Shows cross-reactivity with smooth muscle metavinculin.

Immunogen

purified human vinculin from uterus.

Application

Anti-Vinculin antibody, Mouse monoclonal has been used in
  • immunoblotting
  • immunofluorescence staining
  • immunocytochemistry
  • immunohistochemistry and enzyme linked immunosorbent assay (ELISA).

Biochem/physiol Actions

Vinculin functions as one of several interacting proteins involved in anchoring F-actin to the membrane. It has been shown that a sequence of molecular interactions might be involved in the transmembrane assembly of adhesion plaques. vinculin represents a key element in the transmembrane linkage of the extracellular matrix to the cytoplasmic microfilament system. In many cell types undergoing viral transformation, vinculin becomes redistributed to rosettes or podosomes.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loredana Pellegrino et al.
Nucleic acids research, 41(10), 5400-5412 (2013-04-13)
Uncontrolled cell proliferation and cytoskeletal remodeling are responsible for tumor development and ultimately metastasis. A number of studies have implicated microRNAs in the regulation of cancer cell invasion and migration. Here, we show that miR-23b regulates focal adhesion, cell spreading
Functional importance of Dicer protein in the adaptive cellular response to hypoxia
Ho JJD, et al.
Test, 287(34), 29003-29020 (2012)
Biomimetic calcium carbonate concentric microgrooves with tunable widths for promoting MC3T3-E1 cell functions
Wu X and Wang S
Advanced Helathcare Materials, 2(2), 326-333 (2013)
Enhancing integrin alpha1 inserted (I) domain affinity to ligand potentiates integrin alpha1beta1-mediated down-regulation of collagen synthesis
Shi M, et al.
The Journal of Biological Chemistry, 287(42), 35139-35152 (2012)
Podosomes as smart regulators of cellular adhesion
Spinardi L and Marchisio PC
European Journal of Cell Biology, 85(3-4), 191-194 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service