Skip to Content
Merck
All Photos(5)

Key Documents

V4505

Sigma-Aldrich

Anti-Vinculin Antibody

mouse monoclonal, VIN-11-5

Synonym(s):

Vinculin Antibody Sigma, Anti-VCL

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Monoclonal Anti-Vinculin antibody produced in mouse, clone VIN-11-5, ascites fluid

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

VIN-11-5, monoclonal

mol wt

antigen 116 kDa

species reactivity

bovine, human, chicken, mouse

technique(s)

indirect immunofluorescence: 1:50 using cultured chicken fibroblasts
microarray: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VCL(7414)
mouse ... Vcl(22330)

General description

Monoclonal Anti-Vinculin (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.
The vinculin (VCL) gene containing 22 exons is mapped to human chromosome 10q22.1-q23. The gene codes for a 117kDa cytoskeletal protein present in adherens type cell junctions. VCL is a ubiquitously expressed cell-adhesion protein. The protein is characterized with a globular head and a tail domain, which are interconnected by a short proline-rich sequence, an intramolecular interaction between the head and tail conceal many ligand-binding sites in the protein.

Specificity

The antibody reacts best with cultured chicken fibroblasts. Labeling also may be obtained with bovine, human, or mouse cells.

Immunogen

smooth muscle vinculin from chicken gizzard.

Application

Monoclonal Anti-Vinculin antibody produced in mouse has been used in:
  • western blotting.
  • immunofluorescence.[1]
  • immunocytochemistry.

Biochem/physiol Actions

Vinculin (VCL) is a cytoskeletal protein, which plays a vital role in regulation of cell-to-cell and cell to matrix junctions. VCL facilitates normal embryonic development and cardiac function. Aberration or mutation of the gene leads to cardiomyopathy. VCL also plays a crucial role in cell migration and morphogenesis. VCL might facilitate the binding of integrins and cadherins to the actin cytoskeleton. Vinculin plays an important role in integrin αIIbβ3 signaling and function, which is required for platelet aggregation.
Vinculin is suggested to be a possible link between ends of the bundles of actin filaments and the plasma membrane. It has also been proposed that vinculin may be involved in the transmembrane induction of actin bundle formation.

Other Notes

This product can be found as purified product that was produced using cell culture hybridoma product.
SAB4200729 Anti-Vinculin antibody, Mouse monoclonal
clone VIN-11-5, purified from hybridoma cell culture

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Three-dimensional structure of vinculin bound to actin filaments
Janssen MEW, et al.
Molecular Cell, 21(2), 271-281 (2006)
W Xu et al.
Journal of cell science, 111 ( Pt 11), 1535-1544 (1998-05-15)
Vinculin plays a role in signaling between integrins and the actin cytoskeleton. We reported earlier that F9-derived cells lacking vinculin are less spread, less adhesive, and move two times faster than wild-type F9 cells. Expression of intact vinculin in null
E P Moiseyeva et al.
The Journal of biological chemistry, 268(6), 4318-4325 (1993-02-25)
The human vinculin gene contains 22 exons ranging in size from 71 base pairs (bp) to 303 bp (average 155 bp) with the exception of exon 22 which contains 144 bp of coding sequence and 1848 bp of 3'-untranslated sequence
Qin Yu et al.
Scientific reports, 5, 15245-15245 (2015-10-22)
Hypertension is the most common and lethal risk factor for cardiovascular disease (CVD). Numerous variants have been associated with hypertension, however, most of which failed to get replication due to ethnic differences. In this study, we analyzed associations of 10
Julia Fröbel et al.
Molecular & cellular proteomics : MCP, 12(5), 1272-1280 (2013-02-06)
Bleeding complications are a significant clinical problem in patients with myelodysplastic syndromes even at sufficient platelet counts (>50,000/μl). However, the underlying pathology of this hemorrhagic diathesis is still unknown. Here, we analyzed the platelet proteome of patients with myelodysplastic syndromes

Questions

  1. Hello! Has it been published or predicted if this antibody will react with Drosophila vinculin? Or, could you provide where in the human vinculin this antibody interacts? 

    1 answer
    1. There are no available papers that cite using clone VIN-11-5 with Drosophila vinculin. The specific epitope for this antibody has not been mapped. However, there is a paper that maps the epitope binding area of this antibody clone - JBC 266, 8734-8740. 1991, Fusun Kilic, et al. --> "Hence the epitope lies between cysteines 545 and 737." Please see the link below to review this publication:
      https://www.sciencedirect.com/science/article/pii/S0021925818315072?via%3Dihub

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service