Skip to Content
Merck
All Photos(4)

Documents

V8879

Sigma-Aldrich

Vincristine sulfate salt

95.0-105.0% (HPLC), powder or crystals

Synonym(s):

22-Oxovincaleukoblastine sulfate salt, Leurocristine sulfate salt, VCR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H56N4O10 · H2SO4
CAS Number:
Molecular Weight:
923.04
Beilstein:
3924631
EC Number:
MDL number:
UNSPSC Code:
51111767
PubChem Substance ID:
NACRES:
NA.85

biological source

Streptomyces roseosporus

Quality Level

Assay

95.0-105.0% (HPLC)

form

powder or crystals

storage condition

(Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.)

color

white to light yellow

antibiotic activity spectrum

neoplastics

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

OS(O)(=O)=O.CC[C@]1(O)CC2CN(CCc3c([nH]c4ccccc34)[C@@](C2)(C(=O)OC)c5cc6c(cc5OC)N(C=O)C7[C@](O)([C@H](OC(C)=O)[C@]8(CC)C=CCN9CC[C@]67C89)C(=O)OC)C1

InChI

1S/C46H56N4O10.H2O4S/c1-7-42(55)22-28-23-45(40(53)58-5,36-30(14-18-48(24-28)25-42)29-12-9-10-13-33(29)47-36)32-20-31-34(21-35(32)57-4)50(26-51)38-44(31)16-19-49-17-11-15-43(8-2,37(44)49)39(60-27(3)52)46(38,56)41(54)59-6;1-5(2,3)4/h9-13,15,20-21,26,28,37-39,47,55-56H,7-8,14,16-19,22-25H2,1-6H3;(H2,1,2,3,4)/t28-,37-,38+,39+,42-,43+,44+,45-,46-;/m0./s1

InChI key

AQTQHPDCURKLKT-PNYVAJAMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Vincristine is an antitumor alkaloid isolated from Vinca Rosea. It is used to treat acute leukaemia, malignant lymphoma, Hodgkin′s disease, acute erythraemia, acute panmyelosis, breast cancer, Kaposi′s sarcoma and testicular cancer. Vincristine is widely used in cancer research. It is used to study multi-drug resistance. It is used to inhibit cell cycle progression at M-phase and to inhibit monoamine oxidase (MAO) .

Biochem/physiol Actions

Vincristine is a plant alkaloid that inhibits microtubule assembly by binding tubulin and inducing coiled spiral aggregate formation. It arrests cell cycle in G2/M-phase by blocking mitotic spindle formation. Vincristine triggers Raf-1 activation, phosphorylation of bcl-2-family proteins, induction of p53 expression, and apoptosis in several tumor cell lines. It inhibits VEGF production in leukemia cell lines. It is a substrate for Pgp and CYP3A4. MRP1 transports vincristine in an ATP- and GSH-dependent manner. Vincristine has some immunosuppressant effect. Vincristine may also interfere with amino acid, cyclic AMP, and glutathione metabolism, calmodulin-dependent Ca2+-transport, cellular respiration, and nucleic acid and lipid biosynthesis.

Analysis Note

Freely soluble in water, Soluble in methanol, slightly soluble in ethanol

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Muta. 2 - Repr. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stacey J Baker et al.
Molecular cell, 79(1), 180-190 (2020-07-04)
Rigosertib is a styryl benzyl sulfone that inhibits growth of tumor cells and acts as a RAS mimetic by binding to Ras binding domains of RAS effectors. A recent study attributed rigosertib's mechanism of action to microtubule binding. In that
J Renes et al.
British journal of pharmacology, 126(3), 681-688 (1999-04-03)
The present study was performed to investigate the ability of the multidrug resistance protein (MRPI) to transport different cationic substrates in comparison with MDR1-P-glycoprotein (MDR1). Transport studies were performed with isolated membrane vesicles from in vitro selected multidrug resistant cell
Moutushy Mitra et al.
Investigative ophthalmology & visual science, 52(8), 5441-5448 (2011-05-07)
To explore the possibility of stathmin as an effective therapeutic target and to evaluate the synergistic combination of stathmin RNAi and the antimicrotubule agents paclitaxel and vincristine to retinoblastoma Y79 cells. RNAi-mediated specific inhibition of stathmin expression in Y79 cells
Trisha A Soosay Raj et al.
International journal of nanomedicine, 8, 4361-4369 (2013-11-16)
Vincristine (VCR) is one of the most extensively used cytotoxic compounds in hemato-oncology. VCR is particularly important for the treatment of acute lymphoblastic leukemia (ALL), a disease that accounts for approximately one-third of all childhood cancer diagnoses. VCR's full therapeutic
Richard Delarue et al.
The Lancet. Oncology, 14(6), 525-533 (2013-04-13)
Immunochemotherapy with rituximab and cyclophosphamide, doxorubicin, vincristine, and prednisone (R-CHOP) has become the standard of care for elderly patients with diffuse large B-cell lymphoma. We aimed to ascertain if a dose-dense R-CHOP regimen administered every 2 weeks (R-CHOP14) was superior

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service