Skip to Content
Merck
All Photos(2)

Key Documents

V4888

Sigma-Aldrich

Anti-VSV-G antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Vesicular Stomatitis Virus glycoprotein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.56

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mammals

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 0.5 μg using VSV-G tagged fusion proteins from transfected mammalian cell lysates (tagged fusion proteins from transfected mammalian cell lysates)
indirect immunofluorescence: 1.0 μg/mL using VSV-G tagged fusion proteins in methanol/acetone fixed transiently transfected cells
western blot: 0.1-0.2 μg/mL using VSV-G tagged fusion proteins from transfected mammalian cell lysates

shipped in

dry ice

storage temp.

−20°C

General description

The Anti-VSV-G rabbit antibody reacts with amino- and carboxyl-terminal VSV-G tagged fusion proteins.
Vesicular stomatitis viruses (VSV) belongs to the Vesiculovirus genus of the Rhabdoviridae family. Vesicular stomatitis virus G (VSV G) protein is considered to be the early recognized type III viral fusion protein.

Immunogen

synthetic peptide corresponding to amino acids 497-511 (N-Tyr-Thr-Asp-Ile-Glu-Met-Asn-Arg-Leu-Gly-Lys-C) of the Vesicular Stomatitis Virus glycoprotein (VSV-G), conjugated to maleimide activated KLH via a cysteine residue added at the carboxyl terminus. The antibody recognizes the VSV-G tag at both the C-terminus and N-terminus.

Application

Anti-VSV-G antibody produced in rabbit has been used in western blotting and immunoprecipitation.
Antibody is suitable for immunoblotting, immunoprecipitation, and indirect immunofluorescence.
immunocytochemistry.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Chromatin immunoprecipitation (1 paper)
Western Blotting (1 paper)
Western blotting following immunoprecipitation (1 paper)

Biochem/physiol Actions

Vesicular stomatitis virus G (VSV G) is capable of stimulating cell-cell fusion upon low pH stimulation. The transmembrane region of VSV G protein is highly essential for the virus transition from the hemi-fusion to full fusion state.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide as a preservative.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vesicular stomatitis virus glycoprotein is a determinant of pathogenesis in swine, a natural host
Martinez I, et al.
Journal of virology, 77(14), 8039-8047 (2003)
Periodic fever with aphthous stomatitis, pharyngitis, and cervical adenitis syndrome is associated with a CARD8 variant unable to bind the NLRP3 inflammasome
Cheung MS, et al.
Journal of immunology (Baltimore, Md. : 1950), 198(5), 2063-2069 (2017)
Vesicular stomatitis virus G protein transmembrane region is crucial for the hemi-fusion to full fusion transition
Ci Y, et al.
Scientific Reports, 8(1), 10669-10669 (2018)
A mechanism for Wnt coreceptor activation
Tamai K, et al.
Molecular Cell, 13(1), 149-156 (2004)
Adenomatous polyposis coli (APC) regulates multiple signaling pathways by enhancing glycogen synthase kinase-3 (GSK-3) activity
Valvezan AJ, et al.
The Journal of Biological Chemistry, 287(6), 3823-3832 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service