Skip to Content
Merck
All Photos(1)

Key Documents

SAB3500946

Sigma-Aldrich

Anti-HIV-1 P24 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 24 kDa

species reactivity

virus

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunoblotting: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

P24 is a free HIV capsid antigen. This antigen forms a part of inner shell of the virus. Less amount of P24 antigen in the blood shows high amount of P24 antibody.

Immunogen

Antibody was raised against a recombinant full-length HIV-1 p24 protein .

Application

Anti-HIV-1 P24 antibody has been used in western blotting.

Biochem/physiol Actions

P24 antigen test is usually done to evaluate new drugs.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yimeng Wang et al.
Journal of virology, 91(7) (2017-01-20)
Interferon-inducible transmembrane proteins (IFITMs) inhibit a broad spectrum of viruses, including HIV-1. IFITM proteins deter HIV-1 entry when expressed in target cells and also impair HIV-1 infectivity when expressed in virus producer cells. However, little is known about how viruses
Daniel Y Joh et al.
Advanced healthcare materials, 8(8), e1801177-e1801177 (2019-03-26)
Poly(ethylene glycol) (PEG), a linear polymer known for its "stealth" properties, is commonly used to passivate the surface of biomedical implants and devices, and it is conjugated to biologic drugs to improve their pharmacokinetics. However, its antigenicity is a growing
Keli Chai et al.
Frontiers in microbiology, 12, 675201-675201 (2021-06-08)
Interferon exerts its antiviral activity by stimulating the expression of antiviral proteins. These interferon stimulate genes (ISGs) often target a group of viruses with unique molecular mechanisms. One such ISG is myxovirus resistance B (MxB) that has been reported to
The V3 loop of HIV-1 Env determines viral susceptibility to IFITM3 impairment of viral infectivity.
Wang Y, et al.
Journal of Virology, 91(7), e02441-e02416 (2017)
Effect of HIV-1 Env on SERINC5 antagonism.
Beitari S, et al.
Journal of Virology, 91(4), e02214-e02216 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service