Skip to Content
Merck
All Photos(2)

Documents

UC148

Sigma-Aldrich

6-Hydroxychlorzoxazone

≥98% (HPLC), solid, chlorzoxazone metabolite

Synonym(s):

5-Chloro-6-hydroxy-2(3H)-benzoxazolone, 5-Chloro-6-hydroxybenzoxazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4ClNO3
CAS Number:
Molecular Weight:
185.56
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

product name

6-Hydroxychlorzoxazone, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

solid

color

white to pink

solubility

methanol: soluble

storage temp.

2-8°C

SMILES string

Oc1cc2OC(=O)Nc2cc1Cl

InChI

1S/C7H4ClNO3/c8-3-1-4-6(2-5(3)10)12-7(11)9-4/h1-2,10H,(H,9,11)

InChI key

AGLXDWOTVQZHIQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

6-Hydroxychlorzoxazone has been used for HPLC-based metabolic assays of chlorzoxazone.
6-hydroxy Chlorzoxazone has been used: as a reference standard to monitor substrate depletion or 6-hydroxy chlorzoxazone formation by cytochrome P450 family 2 subfamily E member 1 (CYP2E1)in recombinant human enzyme screening. 6-Hydroxychlorzoxazone has been used for high-performance liquid chromatography (HPLC)-based metabolic assays of chlorzoxazone.
CYP2E1 & 1A2 metabolite of chlorzoxazone.

Biochem/physiol Actions

6-hydroxy Chlorzoxazone is a novel metabolite of chlorzoxazone. It is formed by the hydroxylation of chlorzoxazone by cytochrome P450 family 2 subfamily E member 1 (CYP2E1) enzyme. The determination of formation and clearance of 6-hydroxychlorzoxazone is used as a reliable marker of CYP2E1 metabolic activity.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

6-Hydroxychlorzoxazone is soluble in methanol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro CYP/FMO Reaction Phenotyping
Optimization in Drug Discovery: In Vitro Methods, 137-169 (2014)
C B Eap et al.
Journal of chromatography. B, Biomedical sciences and applications, 705(1), 139-144 (1998-03-14)
A gas chromatographic-mass spectrometric method is presented which allows the determination of chlorzoxazone and 6-hydroxychlorzoxazone after derivatization with the reagent N-tert.-butyldimethylsilyl-N-methyltrifluoroacetamide. No interference was observed from endogenous compounds following the extraction of plasma samples from six different human subjects. The
Drug Metabolism in Chronic Kidney Disease
Chronic Renal Disease, 1035-1051 (2020)
Eva González-Jasso et al.
Toxicology letters, 144(1), 55-67 (2003-08-16)
The inducibility of CYP2E1 was investigated in liver and peripheral lymphocytes of rats treated with benzene (0-10 mmol/kg body weight (bw), daily for 3 days, i.p., or 0 and 5 mmol/kg bw, daily for 14 days, i.p.) or toluene (0
Hao Wang et al.
Pharmaceutical biology, 58(1), 1150-1155 (2020-12-18)
Succinic acid, extracted from amber, is widely used in cardiovascular therapy. The effect of succinic acid on the activity of cytochrome P450 (CYP450) enzymes was investigated in this study. The effect of succinic acid (100 μM) on the activity of eight

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service