Skip to Content
Merck
All Photos(3)

Documents

C4397

Sigma-Aldrich

Chlorzoxazone

Synonym(s):

5-Chloro-2(3H)-benzoxazolone, 5-Chloro-2-hydroxybenzoxazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4ClNO2
CAS Number:
Molecular Weight:
169.57
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (TLC)

form

powder

color

white to off-white

mp

191-192 °C (lit.)

solubility

DMSO: 50 mg/mL, clear

originator

Johnson & Johnson

storage temp.

room temp

SMILES string

Clc1ccc2OC(=O)Nc2c1

InChI

1S/C7H4ClNO2/c8-4-1-2-6-5(3-4)9-7(10)11-6/h1-3H,(H,9,10)

InChI key

TZFWDZFKRBELIQ-UHFFFAOYSA-N

Gene Information

human ... KCNN4(3783)

Looking for similar products? Visit Product Comparison Guide

Application

Chlorzoxazone may be used:
  • as a calcium-activated potassium (KCa) channel activator in patch-clamp electrophysiology to test its effect on dendritic hyperexcitability
  • as a substrate for cytochrome P450 2E1 (CYP2E1) enzyme in cultured human hepatocytes
  • as a benzimidazolone compound to test its effect on epithelial sodium (Na+) transport

Biochem/physiol Actions

Chlorzoxazone inhibits the multisynaptic reflex arcs primarily in the spinal cord and brain subcortical areas. It is an activator of calcium activated potassium channels and may contribute to the aortic artery relaxation in mice. Chlorzoxazone also activates the calcium-activated potassium (KCa) channel and small conductance calcium-activated potassium channels (SK). It is metabolized by cytochrome P450 2E1 (CYP2E1) and serves as a probe for monitoring this enzyme function.
Chlorzoxazone is a skeletal muscle relaxant.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B J Powers et al.
Archives of internal medicine, 146(6), 1183-1186 (1986-06-01)
A drug rechallenge proved chlorzoxazone to be hepatotoxic in a patient who had been treated with a combination of it and acetaminophen (Parafon Forte) for several months. Failure to demonstrate a toxic response to acetaminophen coupled with a dramatic response
Sanjeev Kumar et al.
International journal of applied & basic medical research, 4(2), 101-105 (2014-08-22)
The fixed dose combinations (FDCs) of muscle relaxants, non-steroidal anti-inflammatory drugs and paracetamol are commonly prescribed in the treatment of acute lower backache. The present study was undertaken with the aim of comparing the efficacy and safety of FDCs of
C A Syme et al.
American journal of physiology. Cell physiology, 278(3), C570-C581 (2000-03-11)
We previously characterized 1-ethyl-2-benzimidazolinone (1-EBIO), as well as the clinically useful benzoxazoles, chlorzoxazone (CZ), and zoxazolamine (ZOX), as pharmacological activators of the intermediate-conductance Ca(2+)-activated K(+) channel, hIK1. The mechanism of activation of hIK1, as well as the highly homologous small-conductance
De-Li Dong et al.
European journal of pharmacology, 545(2-3), 161-166 (2006-07-25)
Chlorzoxazone has been reported to activate the intermediate-conductance, Ca(2+)-activated K(+) channels in aortic endothelial cells and to relax the artery. The aim of the present study was to characterize the chlorzoxazone-induced relaxation of rat thoracic artery. Chlorzoxazone did not affect
Karina Alviña et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 30(21), 7249-7257 (2010-05-28)
Episodic ataxia type-2 (EA2) is an inherited movement disorder caused by mutations in the gene encoding the Ca(v)2.1alpha1 subunit of the P/Q-type voltage-gated calcium channel that result in an overall reduction in the P/Q-type calcium current. A consequence of these

Articles

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service