Skip to Content
Merck
All Photos(5)

Key Documents

U4625

Sigma-Aldrich

Uridine 5′-diphosphoglucose disodium salt hydrate from Saccharomyces cerevisiae

≥98%

Synonym(s):

UDP-Glc hydrate, UDPG hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N2Na2O17P2 · xH2O
CAS Number:
Molecular Weight:
610.27 (anhydrous basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

Saccharomyces cerevisiae

Quality Level

Assay

≥98%

form

powder

storage temp.

−20°C

SMILES string

O.[Na+].[Na+].OC[C@H]1O[C@H](OP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N3C=CC(=O)NC3=O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C15H24N2O17P2.2Na.H2O/c18-3-5-8(20)10(22)12(24)14(32-5)33-36(28,29)34-35(26,27)30-4-6-9(21)11(23)13(31-6)17-2-1-7(19)16-15(17)25;;;/h1-2,5-6,8-14,18,20-24H,3-4H2,(H,26,27)(H,28,29)(H,16,19,25);;;1H2/q;2*+1;/p-2/t5-,6-,8-,9-,10+,11-,12-,13-,14-;;;/m1.../s1

InChI key

SRHFBUUOPANXBG-WSIJJEQHSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Uridine diphosphate glucose (UDP-glucose) is a glycogen precursor. The transfer of glucose from UDP-glucose to growing glycogen chain is catalyzed by the enzyme glycogen synthase. The enzyme uridine diphosphate galactose-4-epimerase catalyzes the formation of UDP-glucose from UDP-galactose.

Application

Uridine 5′-diphosphoglucose disodium salt hydrate from Saccharomyces cerevisiae has been used as a substrate
  • for the PpIRX10 enzyme activity from Arabidopsis thaliana
  • in radiometric assay of glycosyltransferases (GTs)
  • in uridine diphosphate glucose (UDP-Glc) 4-epimerase assay in Arabidopsis thaliana samples

Biochem/physiol Actions

Uridine diphosphate glucose (UDP-glucose) is an essential mediator of the cellular metabolic pathways. In yeast, the high levels of trehalose and glycogen favor increased levels of UDP-glucose.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hypoglycemia as a Pathological Result in Medical Praxis
Type 1 Diabetes Complications (2011)
Distinct properties of the five UDP-D-glucose/UDP-D-galactose 4-epimerase isoforms of Arabidopsis thaliana
Barber C, et al.
The Journal of Biological Chemistry, 281(25), 17276-17285 (2006)
Fariborz Parandeh et al.
The Journal of biological chemistry, 295(45), 15245-15252 (2020-08-29)
Type-2 diabetes (T2D) is a global disease caused by the inability of pancreatic β-cells to secrete adequate insulin. However, the molecular mechanisms underlying the failure of β-cells to respond to glucose in T2D remains unknown. Here, we investigated the relative
A quaternary mechanism enables the complex biological functions of octameric human UDP-glucose pyrophosphorylase, a key enzyme in cell metabolism
Fuhring J, et al.
Scientific Reports, 5, 9618-9618 (2015)
Radiometric and spectrophotometric in vitro assays of glycosyltransferases involved in plant cell wall carbohydrate biosynthesis
Brown C, et al.
Nature Protocols, 7(9), 1634-1634 (2012)

Articles

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service