Skip to Content
Merck
All Photos(2)

Key Documents

G5131

Sigma-Aldrich

Guanosine 5′-diphospho-D-mannose sodium salt from Saccharomyces cerevisiae

Type I, ≥97% (HPLC)

Synonym(s):

GDP-Man, GDP-mannose

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C16H23N5O16P2Na2
CAS Number:
Molecular Weight:
649.30
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Pricing and availability is not currently available.

type

Type I

Quality Level

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

[Na].NC1=NC(=O)c2ncn(C3OC(COP(O)(=O)OP(O)(=O)OC4OC(CO)C(O)C(O)C4O)C(O)C3O)c2N1

InChI

1S/C16H25N5O16P2.Na.H/c17-16-19-12-6(13(28)20-16)18-3-21(12)14-10(26)8(24)5(34-14)2-33-38(29,30)37-39(31,32)36-15-11(27)9(25)7(23)4(1-22)35-15;;/h3-5,7-11,14-15,22-27H,1-2H2,(H,29,30)(H,31,32)(H3,17,19,20,28);;

InChI key

MEXITZOHWLXZKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Guanosine 5′-diphospho-D-mannose (GDP-D-mannose) is present in high cytosolic levels in Saccharomyces cerevisiae. It is crucial for protein mannosylation.[1] GDP-D-mannose is an important precursor to produce GDP-l-fucose.[1]

Application

Guanosine 5′-diphospho-D-mannose sodium salt from Saccharomyces cerevisiae has been used as a nucleotide sugar in transglycosylation enzymatic assay and hydrolysis based catalytic reactions with S-glycosyltransferases (SunS).[2]
Guanosine 5′-diphospho-D-mannose sodium salt has been used as a component of ATP regeneration system for in vitro vesicle formation from Saccharomyces cerevisiae microsomes.[3] It has also been used as a component of the buffer for mannosyl transferase activity assay.[4]

Biochem/physiol Actions

Guanosine diphosphate mannose (GDP-mannose) is a nucleotide sugar substrate for glycosyltransferase (mannosylation) reactions mediated by mannosyltransferases.[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct investigation of the Aspergillus GDP-mannose transporter by STD NMR spectroscopy.
Andrea Maggioni et al.
Chembiochem : a European journal of chemical biology, 12(16), 2421-2425 (2011-09-29)
Two KTR Mannosyltransferases Are Responsible for the Biosynthesis of Cell Wall Mannans and Control Polarized Growth in Aspergillus fumigatus
Henry C, et al.
mBio, 10(1), e02647-e02618 (2019)
Lysophospholipids facilitate COPII vesicle formation
Melero A, et al.
Current Biology, 28(12), 1950-1958 (2018)
The coronary club.
M M Gilliland
Supervisor nurse, 10(3), 50-51 (1979-03-01)
N Smirnoff
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 355(1402), 1455-1464 (2000-12-29)
Ascorbate (vitamin C) can reach very high concentrations in chloroplasts (20-300 mM). The pool size in leaves and chloroplasts increases during acclimation to high light intensity and the highest concentrations recorded are in high alpine plants. Multiple functions for ascorbate

Articles

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

LC-MS/MS method quantifies similar polar nucleotide activated sugars using Supel™ Carbon LC column for simultaneous analysis.

LC-MS/MS method quantifies similar polar nucleotide activated sugars using Supel™ Carbon LC column for simultaneous analysis.

See All

Questions

  1. How to dissolve Guanosine 5′-diphospho-D-mannose sodium salt? 

    1 answer
    1. This product is soluble at 50 mg/mL in water.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service