Skip to Content
Merck
All Photos(3)

Key Documents

U0251

Sigma-Aldrich

Urease from Canavalia ensiformis (Jack bean)

Type C-3, powder, ≥600,000 units/g solid

Synonym(s):

Jack bean urease, Urea amidohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

Canavalia ensiformis

Quality Level

type

Type C-3

form

powder

specific activity

≥600,000 units/g solid

mol wt

~544620 Da

purified by

crystallization

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Subunit molecular weight: ~90,770
Composed of six subunits with total molecular weight: ~544,620
Urease is a nickel-dependent metalloenzyme , found in various plants, bacteria, fungi, and algae. It contains two distinct subunits-UreA(26.5 kDa) and UreB (60.3–61.6 kDa).

Biochem/physiol Actions

Urease aids in the bioavailability of nitrogen in plants. It is implicated in defense mechanism and exhibits insecticidal activity. In addition, urease exhibits antifungal activity by retarding growth and affecting membrane integrity of filamentous fungi and yeasts.
Urease was shown to induce vacuolation in Hela cells, an effect dependent on the ability of urease to convert urea to ammonia. [1]

Unit Definition

One micromolar unit will liberate 1.0 μmole of NH3 from urea per min at pH 7.0 at 25 °C. It is equivalent to 1.0 I.U. or 0.054 Sumner unit (1.0 mg ammonia nitrogen in 5 minutes at pH 7.0 at 20 °C)

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T L Cover et al.
Infection and immunity, 59(4), 1264-1270 (1991-04-01)
Concentrated broth culture supernatants from 50 to 60% of Helicobacter pylori strains induce eukaryotic cell vacuolation in vitro. A quantitative assay for cell vacuolation was developed on the basis of the rapid uptake of visibly vacuolated HeLa cells was significantly
Barbara Krajewska et al.
Plant physiology and biochemistry : PPB, 43(7), 651-658 (2005-07-19)
A pH-variation study of jack bean (Canavalia ensiformis) urease steady-state kinetic parameters and of the inhibition constant of boric acid, a urease competitive inhibitor, was performed using both noninhibitory organic (MES, HEPES and CHES) and inhibitory inorganic (phosphate) buffers, in
J Ryan Feathers et al.
Journal of structural biology: X, 5, 100047-100047 (2021-04-06)
The resolution of cryo-EM reconstructions is fundamentally limited by the Nyquist frequency, which is half the sampling frequency of the detector and depends upon the magnification used. In principle, super-resolution imaging should enable reconstructions to surpass the physical Nyquist limit
J Jessy et al.
The Biochemical journal, 277 ( Pt 3), 693-696 (1991-08-01)
Recent studies showed that hyperammonaemia caused many of the metabolic changes in portacaval-shunted rats, a model of hepatic encephalopathy. These changes included a depression in the cerebral metabolic rate of glucose (CMRGlc), an indication of decreased brain function. 2. The
Wu-Chen Yang et al.
Vaccine, 31(8), 1204-1209 (2013-01-12)
An epitope-based vaccine is a promising option for treating Helicobacter pylori (H. pylori) infection. Epitope mapping is the first step in designing an epitope-based vaccine. A pivotal role of CD4(+) T cells in protection against H. pylori has been accepted

Articles

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service