Skip to Content
Merck
All Photos(1)

Documents

T9576

Sigma-Aldrich

Picropodophyllotoxin

≥96% (HPLC), powder

Synonym(s):

(5R,5aS,8aR,9R)-9-Hydroxy-5,8,8a,9-tetrahydro-5-(3,4,5-trimethoxyphenyl)furo[3′,4′:6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, NSC 36407, PPP, Picropodophyllin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22O8
CAS Number:
Molecular Weight:
414.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥96% (HPLC)

form

powder

color

white

solubility

DMSO: >5 mg/mL

storage temp.

2-8°C

SMILES string

COc1cc(cc(OC)c1OC)[C@H]2C3C(COC3=O)[C@@H](O)c4cc5OCOc5cc24

InChI

1S/C22H22O8/c1-25-16-4-10(5-17(26-2)21(16)27-3)18-11-6-14-15(30-9-29-14)7-12(11)20(23)13-8-28-22(24)19(13)18/h4-7,13,18-20,23H,8-9H2,1-3H3/t13-,18+,19+,20-/m0/s1

InChI key

YJGVMLPVUAXIQN-HAEOHBJNSA-N

Gene Information

human ... IGF1R(3480)

Biochem/physiol Actions

Picropodophyllotoxin (PPP), an epimer of podophyllotoxin (PPT), has been reported to target insulin-like growth factor-I receptor (IGF-IR) and mediate anticancer functions. However, a study in 2007, has reported that PPP can induce G2-M cell cycle arrest even in IGF-IR deficient cells. Hence, the role of PPP in IGF-IR-mediated anticancer functions is debatable.
Picropodophyllotoxin is an insulin-like growth factor-I (IGF-I) receptor kinase inhibitor.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the InsR page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Quality

May contain 2-3% ethanol (w/w)

Preparation Note

Picropodophyllotoxin is soluble in DMSO at a concentration that is greater than 5 mg/ml.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Strömberg et al.
Blood, 107(2), 669-678 (2005-09-17)
Emerging evidence suggests the insulin-like growth factor-1 receptor (IGF-1R) to be an important mediator of tumor-cell survival and resistance to cytotoxic therapy in multiple myeloma (MM). Recently, members of the cyclolignan family have been shown to selectively inhibit the receptor
Vladimir M Shkolnikov et al.
The Lancet. Public health, 4(4), e181-e188 (2019-04-08)
Since 2005, Russia has made substantial progress, experiencing an almost doubling of per-capita gross domestic product by purchasing power parity (GDP [PPP]) to US$24 800 and witnessing a 6-year increase in life expectancy, reaching 71·4 years by 2015. Even greater gains
Brenden Jongman et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(18), E2271-E2280 (2015-04-23)
The global impacts of river floods are substantial and rising. Effective adaptation to the increasing risks requires an in-depth understanding of the physical and socioeconomic drivers of risk. Whereas the modeling of flood hazard and exposure has improved greatly, compelling
Amy Ko et al.
PLoS genetics, 13(8), e1006963-e1006963 (2017-08-23)
Pedigrees contain information about the genealogical relationships among individuals and are of fundamental importance in many areas of genetic studies. However, pedigrees are often unknown and must be inferred from genetic data. Despite the importance of pedigree inference, existing methods
Donghua Liu et al.
Nature communications, 9(1), 193-193 (2018-01-18)
Graphene is regarded as a potential surface-enhanced Raman spectroscopy (SERS) substrate. However, the application of graphene quantum dots (GQDs) has had limited success due to material quality. Here, we develop a quasi-equilibrium plasma-enhanced chemical vapor deposition method to produce high-quality

Articles

We offers many products related to InsR for your research needs.

We offers many products related to InsR for your research needs.

We offers many products related to InsR for your research needs.

We offers many products related to InsR for your research needs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service