Skip to Content
Merck
All Photos(2)

Key Documents

T7947

Sigma-Aldrich

(R)-Tomoxetine hydrochloride

solid

Synonym(s):

(R)-N-Methyl-γ-(2-methyl­phenoxy)benzenepropanamine hydrochloride, Atomoxetine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO · HCl
CAS Number:
Molecular Weight:
291.82
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

originator

Eli Lilly

storage temp.

2-8°C

SMILES string

CNCC[C@@H](OC1=CC=CC=C1C)C2=CC=CC=C2.[H]Cl

InChI

1S/C17H21NO.ClH/c1-14-8-6-7-11-16(14)19-17(12-13-18-2)15-9-4-3-5-10-15;/h3-11,17-18H,12-13H2,1-2H3;1H/t17-;/m1./s1

InChI key

LUCXVPAZUDVVBT-UNTBIKODSA-N

Gene Information

human ... SLC6A2(6530)

Looking for similar products? Visit Product Comparison Guide

Application

(R)-Tomoxetine hydrochloride has been used as a noradrenaline reuptake inhibitor:
  • to study the role of L-threo-3,4-dihydroxyphenylserine (L-DOPS) in the pathogenesis of Alzheimer′s disease in mice
  • to study its effects on set shifting in rats
  • to study its effects on rat brain as a result of its long-term use

Biochem/physiol Actions

(R)-Tomoxetine hydrochloride is an efficient inhibitor of presynaptic norepinephrine transporters. It also positively regulates the release of acetylcholine in the prefrontal cortex (PFC). (R)-Tomoxetine hydrochloride binds to the serotonin (5-HT) transporter. It is involved in blocking the cortical N-methyl-D-aspartate (NMDA) receptors.. (R)-Tomoxetine hydrochloride exhibits therapeutic effects against attention-deficit/hyperactivity disorder (ADHD) and comorbid oppositional defiant disorder (ODD).
Norepinephrine uptake blocker.

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grazia Dell'Agnello et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 19(11), 822-834 (2009-09-01)
The primary aim of this study was to assess the efficacy of atomoxetine in improving ADHD and ODD symptoms in paediatric patients with ADHD and comorbid oppositional defiant disorder (ODD), non-responders to previous psychological intervention with parent support. This was
Fatma Gür et al.
Biochemical and biophysical research communications, 534, 927-932 (2020-11-05)
Attention Deficit Hyperactivity Disorder (ADHD) is the most common psychiatric disorder reported particularly in children. Long-term use of antipsychotic drugs used in the treatment of ADHD has been shown to exert toxic effects on the brain. However, not enough research
Chi-Yung Shang et al.
Journal of child and adolescent psychopharmacology, 22(5), 353-363 (2012-10-23)
Atomoxetine is efficacious in reducing symptoms of attention- deficit/hyperactivity disorder (ADHD), but its effect on visual memory and attention needs more investigation. This study aimed to assess the effect of atomoxetine on visual memory, attention, and school function in boys
Himanshu P Upadhyaya et al.
Psychopharmacology, 226(2), 189-200 (2013-02-12)
Treatment of attention-deficit/hyperactivity disorder (ADHD) has for many years relied on psychostimulants, particularly various formulations of amphetamines and methylphenidate. These are central nervous system stimulants and are scheduled because of their abuse potential. Atomoxetine (atomoxetine hydrochloride; Strattera®) was approved in
Romano Arcieri et al.
Journal of child and adolescent psychopharmacology, 22(6), 423-431 (2013-01-31)
The purpose of this study was to assess the cardiovascular effects of drugs used for attention-deficit/hyperactivity disorder (ADHD) in children and adolescents treated in community care centers in Italy. This study was an open, prospective, observational study of youth with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service