Skip to Content
Merck
All Photos(1)

Key Documents

T7326

Sigma-Aldrich

Thrombin from bovine plasma

≥60 NIH units/mg protein (biuret)

Synonym(s):

Factor IIa

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

form

lyophilized powder

specific activity

≥60 NIH units/mg protein (biuret)

composition

Protein, ≥50%

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Thrombin is the final coagulation protease in regard to hemostasis, promoting both procoagulant and anticoagulant effects.[1] Thrombin comprises of A-chain and B-chain with the catalytic pocket. The C-terminal regions has a fibrinogen recognition site (FRS) and a heparin binding site (HBS).[2] The main function of thrombin is cleavage of fibrinogen to fibrin.[3] It coordinates the coagulation cascade and coordinates with protease-activated receptors (PARs) to regulate physiological functions. High levels of thrombin causes neurotoxicity in dopaminergic neurons and contribute to the progression of parkinsons disease.[4] Altered thrombin levels modulates coagulation pathway in multiple sclerosis.[5] Patients with coronary artery disease (CAD) show elevated levels of thrombin.[6]

Application

Thrombin from bovine plasma may be used:
  • for the generation of fibrin from fibrinogen embedded in microbubbles[7]
  • for the cleavage of hydrogenase maturation protein (HypF-N) from the nickel resin[8]

as revitalization model to mimic blood fabrication in root canal[9]
Thrombin is used for site specific cleavage of recombinant fusion proteins containing an accessible thrombin recognition site for removal of affinity tags. Thrombin has been used in a study to evaluate coagulation abnormalities in acute liver failure. [10]

Biochem/physiol Actions

Serine protease that selectively cleaves Arg-Gly bonds in fibrinogen to form fibrin and fibrinopeptides A and B.

Unit Definition

Activity is expressed in NIH units obtained by direct comparison to a NIH thrombin reference standard.

Physical form

Lyophilized powder containing sodium chloride and Tris-HCl, pH 7.0

Preparation Note

Traditional thrombin products are activated with bovine brain thromboplastin, whereas T7326 is activated with bovine lung thromboplastin and does not contain any bovine brain products in its preparation.

Analysis Note

Activity is expressed in NIH units obtained by direct comparison to a NIH Thrombin Reference Standard, Lot K.
The NIH assay procedure uses 0.2 mL of diluted plasma (1:1 with saline) as a substrate and 0.1 mL of thrombin sample (stabilized in a 1% buffered albumin solution) based on a modification of the method of Biggs. Only clotting times in the range of 15-25 seconds are used for determining thrombin concentrations.

Other Notes

View more information on thrombin at www.sigma-aldrich.com/enzymeexplorer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fibrin-Targeted Polymerized Shell Microbubbles as Potential Theranostic Agents for Surgical Adhesions
Gormley CA, et al.
Langmuir (2019)
Backbone NMR assignments of HypF-N under conditions generating toxic and non-toxic oligomers
Patel JR, et al.
Biomolecular NMR Assignments, 1-5 (2018)
Increased plasma thrombin potential is associated with stable coronary artery disease: An angiographically-controlled study
Tosi F, et al.
Thrombosis Research, 155, 16-22 (2017)
Banwari Agarwal et al.
Journal of hepatology, 57(4), 780-786 (2012-06-28)
In acute liver failure (ALF), prothrombin time (PT) and its derivative prothrombin time ratio (PTR) are elevated, and are considered predictors of increased bleeding risk. We aimed at determining whether increased PT/PTR reflects the haemostatic potential and bleeding risk in
Thrombin generation correlates with disease duration in multiple sclerosis (MS): Novel insights into the MS-associated prothrombotic state
Parsons MEM, et al.
Multiple Sclerosis Journal, 3(4), 84-84 (2017)

Questions

  1. Dear team I would like to ask about this product T7326 (Thrombin from bovine plasma ) ,1000U. What is the best way of preparation? If I would like to prepare 2.5U/L How should I do that? Regards

    1 answer
    1. Stock solutions can be prepared at a concentration of 100 units/mL in a 0.1% (w/v) BSA solution. Further dilutions can be prepared fresh to the final desired working concentration.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service