Skip to Content
Merck
All Photos(1)

Documents

RECOMT

Sigma-Aldrich

Thrombin CleanCleave Kit

Synonym(s):

Thrombin Cleavage Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.56

form

suspension

mol wt

37 kDa

technique(s)

protein purification: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

The Thrombin CleanCleave Kit contains a 50% suspension fo thrombin-agarose produced by immobilizing bovine thrombin and is designed for cleavage of recombinant fusion proteins

Application

For cleaving a tag from a recombinant fusion protein which contains the thrombin recognition sequence. Thrombin-agarose is compatible with recombinant proteins expressed in various types of expression systems.

Features and Benefits

  • Fast, efficient cleavage in as little as 2 hours
  • Thrombin is covalently bound to agarose for easy removal.
  • The robust cleavage reaction is effective at temperatures from 4 °C to 37 °C and over a wide range of pH and ionic strengths.
  • Cleave tags even in the presence of 0.1% Triton, 1 M urea or 5 mM EDTA
  • Thrombin-agarose is reusable.

Quantity

200 μl of a 50% slurry of thrombin-agarose cleaves >85% of 1 mg of fusion protein.

Legal Information

CleanCleave is a trademark of Sigma-Aldrich Co. LLC
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Kit Components Only

Product No.
Description

  • Thrombin Cleavage Buffer, 10× 10 mL

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Angeli et al.
Journal of enzyme inhibition and medicinal chemistry, 36(1), 758-763 (2021-03-16)
We report the first activation study of the β-class carbonic anhydrase (CA, EC 4.2.1.1) encoded in the genome of the protozoan pathogen Trichomonas vaginalis, TvaCA1. Among 24 amino acid and amine activators investigated, derivatives incorporating a second carboxylic moiety, such
Carlos Alvarado et al.
Molecules (Basel, Switzerland), 24(18) (2019-09-22)
The Focal Adhesion Targeting (FAT) domain of Focal Adhesion Kinase (FAK) is a promising drug target since FAK is overexpressed in many malignancies and promotes cancer cell metastasis. The FAT domain serves as a scaffolding protein, and its interaction with
Jolanta Krucinska et al.
Biochemistry, 58(9), 1188-1197 (2019-02-05)
Enolase is a glycolytic metalloenzyme involved in carbon metabolism. The advantage of targeting enolase lies in its essentiality in many biological processes such as cell wall formation and RNA turnover and as a plasminogen receptor. We initially used a DARTS
Matthew R Reynolds et al.
Biochemistry, 44(5), 1690-1700 (2005-02-03)
Alzheimer's disease (AD) is a progressive amnestic disorder typified by the pathological misfolding and deposition of the microtubule-associated tau protein into neurofibrillary tangles (NFTs). While numerous post-translational modifications influence NFT formation, the molecular mechanisms responsible for tau aggregation remain enigmatic.
Anna-Maj Albertsson et al.
Journal of neuroinflammation, 11, 197-197 (2014-12-04)
Osteopontin (OPN) is a highly phosphorylated sialoprotein and a soluble cytokine that is widely expressed in a variety of tissues, including the brain. OPN and OPN-derived peptides have been suggested to have potential neuroprotective effects against ischemic brain injury, but

Articles

This page shows how to cleave and purify GST-tagged proteins bound to GSTrap from Cytiva.

Protocols

This page how to remove GST tags by enzymatic cleavage using Cytiva products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service