Skip to Content
Merck
All Photos(4)

Key Documents

T5816

Sigma-Aldrich

Tricine

BioPerformance Certified, suitable for cell culture, ≥99% (titration)

Synonym(s):

2-[[1,3-Dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid, N-[Tris(hydroxymethyl)methyl]glycine, N-[Tris(hydroxymethyl)methyl]glycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2)3CNHCH2CO2H
CAS Number:
Molecular Weight:
179.17
Beilstein:
1937804
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Quality Level

grade

BioPerformance Certified

Assay

≥99% (titration)

form

crystalline powder

storage condition

dry at room temperature

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin and total aerobic microbial count, tested

color

white

useful pH range

7.4-8.8

pKa (25 °C)

8.1

mp

187 °C

cation traces

heavy metals (as Pb): ≤5 ppm

absorption

≤0.02 at 290 at 20%

application(s)

diagnostic assay manufacturing
general analytical
life science and biopharma
sample preparation

SMILES string

OCC(CO)(CO)NCC(O)=O

InChI

1S/C6H13NO5/c8-2-6(3-9,4-10)7-1-5(11)12/h7-10H,1-4H2,(H,11,12)

InChI key

SEQKRHFRPICQDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tricine is a zwitterionic biological buffer commonly used in enzyme assays, electrophoresis, and cell culture research. It has a higher negative charge compared to glycine, which enables it to migrate more rapidly. Tricine′s high ionic strength has the ability to cause more ion movement and less protein movement, resulting in the separation of low molecular weight proteins in lower percent acrylamide gels. It has been observed that Tricine is efficient in the separation of proteins ranging from 1 to 100 kDa through electrophoresis. According to a study, the 25 mmol/L Tricine buffer proved to be the most effective among ten other buffers tested for ATP assays using firefly luciferase. Further, Tricine is also used in capillary zone electrophoresis, high performance liquid chromatography and ion exchange chromatography.

Application

Tricine serves as a versatile buffering agent suitable for separating low molecular weight proteins in lower percent acrylamide gels, has demonstrated efficacy in ATP assays employing firefly luciferase, and has shown potential in scavenging hydroxyl radicals during the investigation of radiation-induced membrane damage. Additionally, it has played a role in isolating and purifying ribosomes engaged in the translation of specific mRNA and containing specific peptidyl-tRNA molecules. Tricine has also been employed in the synthesis of RNA oligonucleotides and the fabrication of high-density DNA microarrays, enabling the exploration of mechanisms underlying nucleic acid·ligand interactions and library preparation through photolithography.
Buffer component for separation of low molecular weight peptides.

Features and Benefits

  • Suitable as a Buffer component, for Electrophoresis and Protein separation
  • Effective Buffering from pH 7.4-8.8 (25 °C) with a pKa of 8.1 (25 °C)
  • Tested for Endotoxins and Total Aerobic Microbial Count
  • Tested to confirm low levels of heavy metal contamination, ensuring suitability for various applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Nitin Shirole et al.
Journal of visualized experiments : JoVE, (48)(48), doi:10-doi:10 (2011-03-16)
Recently, structural and biochemical studies have detailed many of the molecular events that occur in the ribosome during inhibition of protein synthesis by antibiotics and during nascent polypeptide synthesis. Some of these antibiotics, and regulatory nascent polypeptides mostly in the
Yi Zhou et al.
Nature, 607(7919), 527-533 (2022-07-07)
Immature dentate granule cells (imGCs) arising from adult hippocampal neurogenesis contribute to plasticity and unique brain functions in rodents1,2 and are dysregulated in multiple human neurological disorders3-5. Little is known about the molecular characteristics of adult human hippocampal imGCs, and
Thierry Rabilloud
Journal of proteomics, 73(8), 1562-1572 (2010-04-17)
Electrophoretic separations of proteins are widely used in proteomic analyses, and rely heavily on SDS electrophoresis. This mode of separation is almost exclusively used when a single dimension separation is performed, and generally represents the second dimension of two-dimensional separations.
Arpita Gantayet et al.
Biofouling, 29(1), 77-85 (2012-12-06)
The freshwater zebra mussel (Dreissena polymorpha) is a notorious biofouling organism. It adheres to a variety of substrata underwater by means of a proteinaceous structure called the byssus, which consists of a number of threads with adhesive plaques at the
Bin Deng et al.
PloS one, 9(7), e102362-e102362 (2014-07-19)
Maresins are a new family of anti-inflammatory and pro-resolving lipid mediators biosynthesized from docosahexaenoic acid (DHA) by macrophages. Here we identified a novel pro-resolving product, 13R,14S-dihydroxy-docosahexaenoic acid (13R,14S-diHDHA), produced by human macrophages. PCR mapping of 12-lipoxygenase (12-LOX) mRNA sequence in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service