Skip to Content
Merck
All Photos(4)

Documents

T5141

Sigma-Aldrich

Glyceryl trimyristate

≥99%

Synonym(s):

TG(14:0/14:0/14:0), 1,2,3-Tritetradecanoylglycerol, Glycerol trimyristate, Trimyristin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)12COOCH2]2CHOCO(CH2)12CH3
CAS Number:
Molecular Weight:
723.16
Beilstein:
1718242
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

plant (palm)

Assay

≥99%

form

powder

mp

56-57 °C (lit.)

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

SMILES string

CCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCC

InChI

1S/C45H86O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-43(46)49-40-42(51-45(48)39-36-33-30-27-24-21-18-15-12-9-6-3)41-50-44(47)38-35-32-29-26-23-20-17-14-11-8-5-2/h42H,4-41H2,1-3H3

InChI key

DUXYWXYOBMKGIN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maximilian Borgmeyer et al.
Cell reports, 37(1), 109797-109797 (2021-10-06)
Membrane lipids and their metabolism have key functions in neurotransmission. Here we provide a quantitative lipid inventory of mouse and rat synaptic junctions. To this end, we developed a multiomics extraction and analysis workflow to probe the interplay of proteins
W Vogelhuber et al.
Pharmaceutical development and technology, 8(1), 71-79 (2003-04-01)
Matrices made of glyceryl trimyristate as a bioerodible and biocompatible material were manufactured by compression in dimensions that would still allow an application via injection. Pyranine, as a low molecular hydrophilic compound with a low detection limit, and tetramethylrhodamine labeled
S Martins et al.
Journal of biomedical nanotechnology, 5(1), 76-83 (2010-01-09)
Solid lipid nanoparticles (SLN) composed of trimyristin (solid lipid) and poloxamer 407 (surfactant) were prepared by a w/o/w emulsion technique for the incorporation of Salmon calcitonin, and further explored as protein carriers for oral delivery. Trimyristin SLN showed a mean
Carsten Olbrich et al.
International journal of pharmaceutics, 237(1-2), 119-128 (2002-04-17)
In vivo drug release from solid lipid nanoparticles (SLN) takes place by diffusion and degradation of the lipid matrix. SLN with different degree of crystallinity were prepared to study the effect of crystallinity on the degradation velocity. These SLN were
C Thies et al.
Journal of microencapsulation, 20(1), 87-96 (2003-01-10)
A novel solvent-free particle coating process, based on the solvent properties of supercritical fluids (SCF) is described. It consists of dissolving one or more coating materials in supercritical CO(2) and then adjusting T/P conditions in the autoclave so that the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service