Skip to Content
Merck
All Photos(2)

Key Documents

T9517

Sigma-Aldrich

Glyceryl trilinoleate

≥98% (TLC), liquid

Synonym(s):

TG(18:2(9Z,12Z)/18:2(9Z,12Z)/18:2(9Z,12Z)), 1,2,3-Tri-(cis,cis-9,12-octadecadienoyl)glycerol, 1,2,3-Trilinoleoylglycerol, Glycerol trilinoleate, Trilinolein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H98O6
CAS Number:
Molecular Weight:
879.38
Beilstein:
1718698
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥98% (TLC)

form

liquid

density

0.925 g/mL at 20 °C (lit.)

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C/C\C=C/CCCCC)OC(=O)CCCCCCC\C=C/C\C=C/CCCCC

InChI

1S/C57H98O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h16-21,25-30,54H,4-15,22-24,31-53H2,1-3H3/b19-16-,20-17-,21-18-,28-25-,29-26-,30-27-

InChI key

HBOQXIRUPVQLKX-BBWANDEASA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Gum Arabic-Stabilized Ferric Oxyhydroxide Nanoparticles for Efficient and Targeted Intestinal Delivery of Bioavailable Iron.: This research explores the use of glyceryl trilinoleate in stabilizing nanoparticles for targeted iron delivery, highlighting its potential in improving bioavailability and therapeutic efficiency (Li et al., 2023).

Packaging

Sealed ampule.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming-Jun Gao et al.
The Plant cell, 21(1), 54-71 (2009-01-22)
The seed maturation program is repressed during germination and seedling development so that embryonic genes are not expressed in vegetative organs. Here, we describe a regulator that represses the expression of embryonic seed maturation genes in vegetative tissues. ASIL1 (for
Paul Chan et al.
Acta pharmacologica Sinica, 23(12), 1157-1162 (2002-12-06)
Trilinolein is a triacylglycerol purified from a commonly used traditional Chinese medicine Panax notoginseng. Trilinolein has been reported to provide a number of beneficial effects including reducing thrombogenicity and arrhythmias and increasing erythrocyte deformability. Additionally, trilinolein has been reported to
Ju-Chi Liu et al.
Journal of biomedical science, 11(1), 11-18 (2004-01-20)
The myocardial protective effects of trilinolein, isolated from the traditional Chinese herb Sanchi (Panax notoginseng), are thought to be related to its antioxidant activity. However, the intracellular mechanism underlying the protective effect of trilinolein in the heart remains unclear. In
M A Fuller et al.
Archives of biochemistry and biophysics, 388(1), 146-154 (2001-05-22)
In soybean (Glycine max L.) vegetative tissue at least five lipoxygenase isozymes are present. Four of these proteins have been localized to the paraveinal mesophyll, a layer of cells that is thought to function in assimilate partitioning. In order to
Chandra Durgampudi et al.
The American journal of pathology, 184(6), 1773-1784 (2014-05-24)
Obese patients have worse outcomes during acute pancreatitis (AP). Previous animal models of AP have found worse outcomes in obese rodents who may have a baseline proinflammatory state. Our aim was to study the role of acute lipolytic generation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service