Skip to Content
Merck
All Photos(4)

Key Documents

T4510

Sigma-Aldrich

Thymidine 5′-monophosphate p-nitrophenyl ester sodium salt

chromogenic, ≥98% (HPLC), powder

Synonym(s):

p-Nitrophenyl-5V-thymidine-monophosphate, p-Nph-5V-TMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N3O10PNa
CAS Number:
Molecular Weight:
466.29
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

Thymidine 5′-monophosphate p-nitrophenyl ester sodium salt, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

solubility

water: 50 mg/mL

storage temp.

−20°C

SMILES string

CC(C(N1)=O)=CN([C@H]2C[C@H](O)[C@@H](COP(O)(OC(C=C3)=CC=C3[N+]([O-])=O)=O)O2)C1=O.[Na]

InChI

1S/C16H18N3O10P.Na.H/c1-9-7-18(16(22)17-15(9)21)14-6-12(20)13(28-14)8-27-30(25,26)29-11-4-2-10(3-5-11)19(23)24;;/h2-5,7,12-14,20H,6,8H2,1H3,(H,25,26)(H,17,21,22);;

InChI key

PHNRUCBIIRECII-UHFFFAOYSA-N

Application

Thymidine 5′-monophosphate p-nitrophenyl ester sodium salt has been used as a substrate for assessing phosphodiesterase activity. It is also been used as a nucleotide analog in the autotaxin inhibition assay.

Biochem/physiol Actions

Thymidine 5′-monophosphate p-nitrophenyl ester sodium salt is an artificial substrate marker for 5′-nucleotide phosphodiesterase enzyme activity. It is also used as a substrate for autotaxin (ATX, NPP2), which is a member of the ecto-nucleotide pyrophosphatase/phosphodiesterase (NPP) enzyme family.

Substrates

A sensitive chromogenic substrate for venom phosphodiesterase. It is not hydrolyzed by bovine spleen phosphodiesterase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adenosine A2A receptor agonist (CGS-21680) prevents endotoxin-induced effects on nucleotidase activities in mouse lymphocytes.
Vuaden FC, et al.
European Journal of Pharmacology, 651(1-3), 212-217 (2011)
Soluble NTPDase: an additional system of nucleotide hydrolysis in rat blood serum.
Oses JP, et al.
Life Sciences, 74(26), 3275-3284 (2004)
Paul R Stabach et al.
Clinical and translational science, 14(1), 362-372 (2020-10-17)
Enzyme replacement with ectonucleotide pyrophosphatase phospodiesterase-1 (ENPP1) eliminates mortality in a murine model of the lethal calcification disorder generalized arterial calcification of infancy. We used protein engineering, glycan optimization, and a novel biomanufacturing platform to enhance potency by using a
Characterization of non-lipid autotaxin inhibitors.
Hoeglund AB, et al.
Bioorganic & Medicinal Chemistry, 18(2), 769-776 (2010)
Ortal Danino et al.
Rheumatology (Oxford, England), 57(8), 1472-1480 (2018-04-25)
Calcium pyrophosphate deposition (CPPD) is associated with osteoarthritis and is the cause of a common inflammatory articular disease. Ecto-nucleotide pyrophosphatase/phosphodiesterase 1 (eNPP1) is the major ecto-pyrophosphatase in chondrocytes and cartilage-derived matrix vesicles (MVs). Thus, eNPP1 is a principle contributor to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service