Skip to Content
Merck
All Photos(1)

Documents

T3955

Sigma-Aldrich

TNP

≥95% (HPLC)

Synonym(s):

N2-(m-Trifluorobenzyl), N6-(p-nitrobenzyl)purine; N6-[(4-nitrophenyl)methyl]-N2-[[3-(trifluoromethyl)phenyl]methyl]-9H-Purine-2,6-diamine; N6-[(4-nitrophenyl)methyl]-N2-[[3-(trifluoromethyl)phenyl]methyl]-1H-Purine-2,6-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16F3N7O2
CAS Number:
Molecular Weight:
443.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

protect from light

color

yellow

solubility

DMSO: ≥10 mg/mL

originator

Novartis

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(CNc2nc(NCc3cccc(c3)C(F)(F)F)nc4[nH]cnc24)cc1

InChI

1S/C20H16F3N7O2/c21-20(22,23)14-3-1-2-13(8-14)10-25-19-28-17(16-18(29-19)27-11-26-16)24-9-12-4-6-15(7-5-12)30(31)32/h1-8,11H,9-10H2,(H3,24,25,26,27,28,29)

InChI key

DDSBPUYZPWNNGH-UHFFFAOYSA-N

Biochem/physiol Actions

TNP is a potent and selective IP6K inhibitor; less selective inhibitor of IP3K. Discovery of TNP as an inositol trisphosphate (IP3) kinase inhibitor was first reported in 2002, However, a recent reference shows that TNP is more potent as an IP6K inhibitor, making it valuable for study of inositol pyrophosphates.

Features and Benefits

This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

TNP is soluble in DMSO at a concentration that is greater than or equal to 10 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Hugh Billings et al.
AIDS research and human retroviruses, 35(9), 842-852 (2019-07-11)
Anti-HIV envelope (Env) antibodies elicit important Fc receptor functions, including FcγRIIIa-mediated natural killer cell killing of opsonized infected targets. How these antibodies evolve during HIV infection and treatment remains poorly understood. We describe changes in anti-HIV Env IgG using longitudinal
Marco Thio et al.
PloS one, 7(7), e40986-e40986 (2012-08-23)
Beside the production of complete immunoglobulins IgG, IgE, IgA, IgM and IgD, consisting of tetrameric heterodimers of immunoglobulin heavy and light chains, B cells also secrete immunoglobulin free light chains (Ig-fLC). Previous studies showed that Ig-fLCs are able to induce
Robert E Burke et al.
American journal of medical quality : the official journal of the American College of Medical Quality, 33(2), 147-153 (2017-07-12)
Veterans are often transferred from "spoke" Veterans Administration (VA) clinics or hospitals to "hub" tertiary VA hospitals for advanced inpatient care, but they face significant barriers to safe transitions home. The Transitions Nurse Program was developed as an intervention to
Rohit Saluja et al.
PloS one, 7(8), e43547-e43547 (2012-08-24)
Mast cell-derived mediators mediate several of the pathological features of asthma. Microbial infections induce asthma exacerbations in which the contribution of mast cells remains incomprehensible. In this study we have investigated the characteristic expression pattern of Toll-like receptors (TLRs) 1-9
Joerg U Eberle et al.
The Journal of investigative dermatology, 139(9), 1957-1965 (2019-03-27)
The mechanisms leading to allergic skin inflammation, such as atopic dermatitis or urticaria, are poorly defined. Here we used a mouse model for IgE-dependent chronic allergic inflammation to study the role of basophils and eosinophils for induction of pathology. FcεRI

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service