Skip to Content
Merck
All Photos(1)

Key Documents

T2705

Sigma-Aldrich

Topotecan hydrochloride hydrate

≥98% (HPLC and enzymatic), powder, topoisomerase I inhibitor

Synonym(s):

9-[(dimethylamino)methyl]-10-hydroxy-(20S)-camptothecin hydrochloride hydrate, NSC-609669 hydrochloride hydrate, SKF-104864A hydrochloride hydrate, hycamptamine hydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H23N3O5 · xHCl · yH2O
CAS Number:
Molecular Weight:
421.45 (anhydrous free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Topotecan hydrochloride hydrate, ≥98% (HPLC and enzymatic)

Quality Level

Assay

≥98% (HPLC and enzymatic)

form

powder

storage condition

desiccated
protect from light

color

yellow

solubility

DMSO: ≥20 mg/mL

originator

GlaxoSmithKline

storage temp.

−20°C

SMILES string

[n]21c(cc5c([c]2=O)COC(=O)[C@]5(O)CC)c3nc4c(cc3C1)c(c(cc4)O)CN(C)C

InChI

1S/C23H23N3O5/c1-4-23(30)16-8-18-20-12(9-26(18)21(28)15(16)11-31-22(23)29)7-13-14(10-25(2)3)19(27)6-5-17(13)24-20/h5-8,27,30H,4,9-11H2,1-3H3/t23-/m0/s1

InChI key

UCFGDBYHRUNTLO-QHCPKHFHSA-N

Gene Information

human ... TOP1MT(116447)

Application

Topotecan has been used as a positive control for the identification and analysis of HIF-1α and VEGF inhibitors in human glioma cells under hypoxic conditions1. It has also been used for in vitro apoptosis assays in PA317 cells2.

Biochem/physiol Actions

Topotecan is a topoisomerase I inhibitor and an apoptosis inducer. It is a potent antineoplastic agent.

Features and Benefits

This compound is a featured product for ADME Tox and Apoptosis research. Discover more featured ADME Tox and Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

This product is soluble in water, however, the concentration has not been determined. Various sources state this chemical can be solubilized in water at greater than 1 mg/mL.
Topotecan hydrochloride hydrate is soluble in DMSO at a concentration that is greater than or equal to 20 mg/ml.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ya-Lin Lu et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(22) (2021-05-26)
Neuron-enriched microRNAs (miRNAs), miR-9/9* and miR-124 (miR-9/9*-124), direct cell fate switching of human fibroblasts to neurons when ectopically expressed by repressing antineurogenic genes. How these miRNAs function after the repression of fibroblast genes for neuronal fate remains unclear. Here, we
Ian F King et al.
Nature, 501(7465), 58-62 (2013-09-03)
Topoisomerases are expressed throughout the developing and adult brain and are mutated in some individuals with autism spectrum disorder (ASD). However, how topoisomerases are mechanistically connected to ASD is unknown. Here we find that topotecan, a topoisomerase 1 (TOP1) inhibitor
Jerec W Ricci et al.
Molecular cancer therapeutics, 15(12), 2853-2862 (2016-09-28)
Chemotherapeutic resistance remains a challenge in the treatment of ovarian carcinoma, especially in recurrent disease. Despite the fact that most patients with newly diagnosed tumors attain complete remission following cytoreductive surgery and chemotherapy, ovarian carcinoma has a recurrence rate that
Andrei Molotkov et al.
Pharmaceutics, 13(3) (2021-04-04)
Glioblastoma (GBM) is the most common primary adult brain malignancy with an extremely poor prognosis and a median survival of fewer than two years. A key reason for this high mortality is that the blood-brain barrier (BBB) significantly restricts systemically
Iben Kümler et al.
Breast cancer research and treatment, 138(2), 347-358 (2013-03-21)
Following treatment with anthracyclines and taxanes, few established options exist for the treatment of metastatic breast cancer (MBC). Although the topoisomerase 1 inhibitors irinotecan, etirinotecan, and topotecan have been used in clinical trials on MBC, the drugs have never been

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service