Skip to Content
Merck
All Photos(3)

Key Documents

C2538

Sigma-Aldrich

Carboplatin

Synonym(s):

cis-Diammine(1,1-cyclobutanedicarboxylato) platinum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C4H6(CO2)2]Pt(NH3)2
CAS Number:
Molecular Weight:
371.25
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

antibiotic activity spectrum

neoplastics

originator

Bristol-Myers Squibb

SMILES string

N.N.O=C1O[Pt]OC(=O)C12CCC2

InChI

1S/C6H8O4.2H3N.Pt/c7-4(8)6(5(9)10)2-1-3-6;;;/h1-3H2,(H,7,8)(H,9,10);2*1H3;/q;;;+2/p-2

InChI key

OLESAACUTLOWQZ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Carboplatin is used to treat testicular, ovarian and breast cancer.

Application

Carboplatin has been used:
  • in chemotherapy-induced thrombocytopenia mouse model and treatment
  • to induce cell death in a large subset of colorectal cancer cells
  • in the evaluation of cell proliferation and chemosensitivity
  • as a positive control for suppressed cell growth

Biochem/physiol Actions

Carboplatin is a platinum-based antineoplastic drug that damages DNA by forming intrastrand cross-links with neighboring guanine residues. Tumors acquire resistance to these drugs through the loss of DNA-mismatch repair (MMR) activity and the resultant decrease in the induction of programmed cell death.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Bristol-Myers Squibb. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Muta. 1B - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joel Shapiro et al.
Annals of surgery, 258(5), 678-688 (2013-10-08)
To gain insight into the exact location of residual esophageal cancer in the esophageal wall and regional lymph nodes after neoadjuvant chemoradiotherapy (nCRT) and to determine the pattern of regression. Data from the recently published chemoradiotherapy for oesophageal cancer followed
Furqan Shaikh et al.
Pediatric blood & cancer, 60(4), 587-592 (2012-09-15)
While cisplatin is considered superior to carboplatin for the treatment of malignant germ cell tumors (MGCTs) in adults, pediatric oncology collaborative groups still remain concerned about the late effects of cisplatin in children. We performed a literature search to identify
Gamma-secretase inhibition combined with platinum compounds enhances cell death in a large subset of colorectal cancer cells
Aleksic T and Feller SM
Cell communication and signaling : CCS, 6(1), 8-8 (2008)
Selective toxicity of rose bengal to ovarian cancer cells in vitro
Koevary SB
International Journal of Physiology, Pathophysiology and Pharmacology, 4(2), 99-99 (2012)
Nikola A Bowden
Cancer letters, 346(2), 163-171 (2014-01-28)
Platinum based therapy is one of the most effectively used chemotherapeutic treatments for cancer. The mechanism of action of platinum compounds is to damage DNA and drive cells into apoptosis. The most commonly used platinum containing agents are cis-diammine-dichloroplatinum (II)]

Articles

DNA damage and repair mechanism is vital for maintaining DNA integrity. Damage to cellular DNA is involved in mutagenesis, the development of cancer among others.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service