Skip to Content
Merck
All Photos(2)

Key Documents

SRP5009

Sigma-Aldrich

CDK1/CyclinB1, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CCNB, CCNB1, CDC2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

12-16 nmol/min·mg

mol wt

~58 kDa (CDK1)
~75 kDa (CyclinB1)

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

General description

The cyclin B1 (CCNB1) gene is mapped to human chromosome 5q13.2. The gene codes for a 62kDa protein containing a classical cyclin box domain. CCNB1 shuttles between the nucleus and cytoplasm until prophase begins. It is a member of the cyclin family and is expressed at low levels in normal cells. CDK1 (cyclin-dependent kinase 1) belongs to the Ser/Thr protein kinase family. The gene encoding it is localized on human chromosome 10q21.

Biochem/physiol Actions

The cyclin B1 (CCNB1) and cyclin-dependent kinase 1 (CDK1) complex plays a crucial role in the regulation of mitosis. Elevated expression of CCNB1 has been observed in various types of human cancers including breast, lung, colon, prostate and head and neck cancers. Thus, this protein can be considered as a potent target for cancer vaccines.In addition, CCNB1 also serves as a prognostic biomarker for estrogen-receptor-positive (ER+) breast cancer. CDK1 (cyclin-dependent kinase 1) controls various mitotic processes like reorganization of the cytoskeleton and segregation of chromosomes. It is essential for the multiplication of mammalian cells.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CCNB1 is a prognostic biomarker for ER+ breast cancer
Kun Ding
Medical Hypotheses, 83(3) (2014)
Expression analysis of mitotic spindle checkpoint genes in breast carcinoma: role of NDC80/HEC1 in early breast tumorigenicity, and a two-gene signature for aneuploidy
Ivan B
Molecular Cancer (2011)
Soraya Játiva et al.
Cellular and molecular life sciences : CMLS, 76(18), 3601-3620 (2019-03-31)
Exit from mitosis and completion of cytokinesis require the inactivation of mitotic cyclin-dependent kinase (Cdk) activity. In budding yeast, Cdc14 phosphatase is a key mitotic regulator that is activated in anaphase to counteract Cdk activity. In metaphase, Cdc14 is kept
Importance of tuberin in carcinogenesis (Review)
Mingwei Jin
Oncology Letters (2017)
Recurrent CDK1 overexpression in laryngeal squamous cell carcinoma
K. Bednarek
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 37(8) (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service