Skip to Content
Merck
All Photos(1)

Key Documents

SRP3181

Sigma-Aldrich

VCAM-I human

recombinant, expressed in HEK 293 cells, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CD106, INCAM-100, MGC108734, MGC99561, VCAM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

0.8-1.0 μg/mL ED50

mol wt

74.1 kDa

packaging

pkg of 50 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... VCAM1(7412)

General description

Vascular cell adhesion molecule 1 (VCAM-1) is a 110kDa cell surface integral membrane glycoprotein that belongs to the Ig (immunoglobulin)-related superfamily of adhesion molecules. The human VCAM-1 gene codes for a 715 amino acid transmembrane glycoprotein containing a 19 amino acid cytoplasmic domain, a 22 amino acid transmembrane domain, and a 674 amino acid extracellular domain. It is expressed on endothelial cells. Recombinant human VCAM-1 is a 110kDa glycoprotein comprising the extracellular domain (673 amino acid residues) of VCAM-1. Monomeric glycosylated VCAM-1 migrates at an apparent molecular weight of approximately 74.1kDa by SDS-PAGE analysis under reducing conditions.

Biochem/physiol Actions

The primary function of VCAM-1 is the mediation of leukocyte-endothelial cell adhesion and signal transduction. VCAM-1 may play a vital role in the development of several diseases, including atherosclerosis and rheumatoid arthritis. The protein has a crucial role in early embryonic development. It also has a role in lymphocyte trafficking and homeostasis of lymphocyte development.

Sequence

FKIETTPESR YLAQIGDSVS LTCSTTGCES PFFSWRTQID SPLNGKVTNE GTTSTLTMNP VSFGNEHSYL CTATCESRKL EKGIQVEIYS FPKDPEIHLS GPLEAGKPIT VKCSVADVYP FDRLEIDLLK GDHLMKSQEF LEDADRKSLE TKSLEVTFTP VIEDIGKVLV CRAKLHIDEM DSVPTVRQAV KELQVYISPK NTVISVNPST KLQEGGSVTM TCSSEGLPAP EIFWSKKLDN GNLQHLSGNA TLTLIAMRME DSGIYVCEGV NLIGKNRKEV ELIVQEKPFT VEISPGPRIA AQIGDSVMLT CSVMGCESPS FSWRTQIDSP LSGKVRSEGT NSTLTLSPVS FENEHSYLCT VTCGHKKLEK GIQVELYSFP RDPEIEMSGG LVNGSSVTVS CKVPSVYPLD RLEIELLKGE TILENIEFLE DTDMKSLENK SLEMTFIPTI EDTGKALVCQ AKLHIDDMEF EPKQRQSTQT LYVNVAPRDT TVLVSPSSIL EEGSSVNMTC LSQGFPAPKI LWSRQLPNGE LQPLSENATL TLISTKMEDS GVYLCEGINQ AGRSRKEVEL IIQVTPKDIK LTAFPSESVK EGDTVIISCT CGNVPETWII LKKKAETGDT VLKSIDGAYT IRKAQLKDAG VYECESKNKV GSQLRSLTLD VQGRENNKDY FSP

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.0.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved monitoring of clinical response in Systemic Lupus Erythematosus by longitudinal trend in soluble vascular cell adhesion molecule-1.
Lewis MJ
Arthritis Research & Therapy, 18, 5-5 (2016)
Elena Galkina et al.
Arteriosclerosis, thrombosis, and vascular biology, 27(11), 2292-2301 (2007-08-04)
Numerous reports document the role of vascular adhesion molecules in the development and progression of atherosclerosis. Recent novel findings in the field of adhesion molecules require an updated summary of current research. In this review, we highlight the role of
VCAM-1+ placenta chorionic villi-derived mesenchymal stem cells display potent pro-angiogenic activity.
Du W
Stem Cell Research & Therapy, 7, 49-49 (2016)
Soluble Vascular Cell Adhesion Molecule-1 (sVCAM-1) Is Elevated in Bronchoalveolar Lavage Fluid of Patients with Acute Respiratory Distress Syndrome.
Attia EF
PLoS ONE, 11(2), e0149687-e0149687 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service