Skip to Content
Merck
All Photos(1)

Key Documents

SRP3169

Sigma-Aldrich

TFF3 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

ITF, TFI, Intestinal trefoil factor, Trefoil Factor 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

1.0-10.0 mg per mL

mol wt

13.2 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TFF3(7033)

General description

Researcharea: Cell Signaling

The trefoil factor peptides (TFF1, TFF2, and TFF3) are secreted in the gastrointestinal tract. They are secreted peptides which are protease-resistant. TFF-3 is expressed by goblet cells and in the uterus, and has also been shown to be expressed in certain cancers, including colorectal, hepatocellular, and in biliary tumors. Recombinant human TFF3 is a homodimeric protein consisting of two 59 amino acid chains, which includes a 40-amino acid trefoil motif containing three conserved intramolecular disulfide bonds.

Application

TFF3 human has been used in surface plasmon resonance(SPR) binding analysis to study the molecular interactions between TFF3 and the aminonitrile compound and its effect on inhibition of TFF3-dependent cancer cell survival.

Biochem/physiol Actions

The trefoil factor peptides (TFF1, TFF2, and TFF3) play an important role in intestinal mucosal maintenance, defense, and repair. TFF3 also promotes airway epithelial cell migration and differentiation. TFF3 may be useful as a molecular marker for certain types of cancer. TFF3 interacts with the CD147 receptor to promote migration, invasion, and proliferation in colorectal cancer (CRC), Indicating its involvement in cancer progression. TFF3 stimulates the replication of pancreatic islet β-cells in humans and rodents, leading to increased proliferation while preserving their functionality. The effects of TFF3 on β-cell proliferation have been associated with the activation of the protein kinase B(Akt) and epidermal growth factor receptor (EGFR) signaling pathways.

Sequence

EEYVGLSANQ CAVPAKDRVD CGYPHVTPKE CNNRGCCFDS RIPGVPWCFK PLQEAECTF

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P-O Lafontaine et al.
Journal francais d'ophtalmologie, 26(10), 1007-1014 (2003-12-24)
Trefoil factor family (TFF) peptides (formerly P-domain peptides; trefoil factor) are small (7-12 kDa) protease-resistant secreted peptides designated pS2 (or TFF1), SP (TFF2) and ITF (TFF3). Human conjunctival goblet cells (GCs) are known to synthesize TFF, but TFF expression by
Shu Q Liu et al.
PloS one, 8(10), e77732-e77732 (2013-11-10)
Cerebral ischemia, while causing neuronal injury, can activate innate neuroprotective mechanisms, minimizing neuronal death. In this report, we demonstrate that experimental cerebral ischemia/reperfusion injury in the mouse causes upregulation of the secretory protein trefoil factor 3 (TFF3) in the hepatocyte
Ting-yi Du et al.
PloS one, 8(11), e80271-e80271 (2013-11-28)
Trefoil factor 3 (TFF3) is a small peptide that plays an important role in mucosal protection, cell proliferation, and cell migration. The aberrant expression of TFF3 is correlated with gastrointestinal inflammation, solid tumors, and other clinical diseases. The objective of
Dale Porter et al.
Molecular cancer research : MCR, 1(5), 362-375 (2003-03-26)
Gene expression patterns in ductal carcinoma in situ (DCIS), and in invasive, and metastatic breast tumors were determined using serial analysis of gene expression (SAGE). We used mRNA in situ hybridization to examine gene expression at the cellular level and
Steen Seier Poulsen et al.
Regulatory peptides, 126(3), 163-171 (2005-01-25)
Peptides of the trefoil factor family (TFF1, TFF2 and TFF3) are cosecreted with mucus from mucus-producing cells in most organ systems and are believed to interact with mucus to form high-viscosity stable gel complexes. In the gastrointestinal tract, they sustain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service