Skip to Content
Merck
All Photos(1)

Key Documents

SML3270

Sigma-Aldrich

QLT0267

≥98% (HPLC)

Synonym(s):

3,5-Diamino-4-(p-methoxyphenyl)hydrazonopyrazole, 4-((4-Methoxyphenyl)diazenyl)-1H-pyrazole-3,5-diamine, ILK-IN-3, KP 15792, KP-15792, KP15792, QLT 0267, QLT-0267

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N6O
CAS Number:
Molecular Weight:
232.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[nH]1nc(c(c1N)\N=N\c2ccc(cc2)OC)N

InChI

1S/C10H12N6O/c1-17-7-4-2-6(3-5-7)13-14-8-9(11)15-16-10(8)12/h2-5H,1H3,(H5,11,12,15,16)/b14-13+

InChI key

QNRNTYHAOBVOKW-BUHFOSPRSA-N

Biochem/physiol Actions

Orally active, potent and selective integrin-linked kinase (ILK) inhibitor with anti-cancer efficacy in cultures and in vivo.
QLT0267 is an orally active, potent and selective integrin-linked kinase (ILK) inhibitor (IC50 = 26 nM; selectivity: >1000-fold over CK2, CSK, DNA-PK, PIM1, PKB/AKT, PKC, >100-fold over ERK1, GSK3β, LCK, PKA, p70S6K, and RSK1/QLT, >10-fold over CDK1/2/5) that inhibits ILK-dependent cellular signaling (IC50 in 2h = 1 μM against 20 nM EGF-induced pAKT Ser473 phosphorylation in Hth74 cells). QLT0267 treatment causes thyroid cancer cell cycle arrest and apoptosis, leading to anti-proliferation efficacy in cultures (IC50 <6 μM; NPA187, DRO, and K4 cell lines) and in mice in vivo (DRO xenografts-derived tumor growth; 50-100 mg/kg via daily p.o.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sofia Nikou et al.
Journal of molecular histology, 51(4), 385-400 (2020-06-28)
Integrin-linked kinase (ILK) forms a heterotrimeric protein complex with PINCH and PARVIN (IPP) in Focal Adhesions (FAs) that acts as a signaling platform between the cell and its microenvironment regulating important cancer-related functions. We aimed to elucidate the role of
Alexandre Ghazi et al.
Oncoimmunology, 10(1), 1940674-1940674 (2021-07-13)
The CMS4 mesenchymal subtype of colorectal cancer (CRC) is associated with poor prognosis and resistance to treatment. The cellular prion protein PrPC is overexpressed in CMS4 tumors and controls the expression of a panel of CMS4-specific genes in CRC cell
Razan Wafai et al.
Breast cancer research : BCR, 22(1), 136-136 (2020-12-06)
Breast cancers acquire aggressive capabilities via epithelial to mesenchymal transition (EMT), in which various integrins/integrin-linked kinase signalling are upregulated. We investigated this in two patient-derived xenografts (PDXs) developed from breast-to-bone metastases, and its functional significance in a breast cancer cell
Masahiro Shiihara et al.
European journal of cancer (Oxford, England : 1990), 148, 239-250 (2021-03-23)
Pancreatobiliary cancer is a highly aggressive tumour with a dismal prognosis. Personalised medicine represents a promising and effective therapeutic approach for this intractable disease. In this study, we aimed to establish a system for identifying and testing genotype-oriented targeted drugs
Rouhollah Mousavizadeh et al.
Scientific reports, 10(1), 12644-12644 (2020-07-30)
Tendons are specialized tissues composed primarily of load-responsive fibroblasts (tenocytes) embedded in a collagen-rich extracellular matrix. Habitual mechanical loading or targeted exercise causes tendon cells to increase the stiffness of the extracellular matrix; this adaptation may occur in part through

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service