Skip to Content
Merck
All Photos(1)

Documents

SRP3039

Sigma-Aldrich

FGF-23 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture

Synonym(s):

Fibroblast Growth Factor-23

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

25.5 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FGF23(8074)

General description

Fibroblast growth factor-23 (FGF-23) is a bone-derived hormone which is secreted by osteocytes and osteoblasts. It is part of the atypical FGF subfamily. The gene encoding it is localized on human chromosome 12p13. Recombinant human FGF-23 is a 25.5kDa globular protein containing 228 amino acid residues.

Biochem/physiol Actions

The FGF family plays a central role during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. Fibroblast growth factor-23, -21 and -19 (FGF-23, FGF-21 and FGF-19) act as circulating hormones and require the participation of a Klotho protein as a co-receptor for their signaling. The signaling receptor for FGF-23, a Klotho-FGFR1 (IIIc) complex, is an essential regulator of the renal sodium phosphate co-transporter and key vitamin D-metabolizing enzymes cytochrome P450 family 27 subfamily B member 1 (CYP27B1) and cytochrome P450 family 24 subfamily A member 1 (CYP24A1). FGF-23 acts in the kidney to regulate phosphate homeostasis and vitamin D metabolism.

Sequence

MYPNASPLLG SSWGGLIHLY TATARNSYHL QIHKNGHVDG APHQTIYSAL MIRSEDAGFV VITGVMSRRY LCMDFRGNIF GSHYFDPENC RFQHQTLENG YDVYHSPQYH FLVSLGRAKR AFLPGMNPPP YSQFLSRRNE IPLIHFNTPI PRRHTRSAED DSERDPLNVL KPRARMTPAP ASCSQELPSA EDNSPMASDP LGVVRGGRVN THAGGTGPEG CRPFAKFI

Physical form

Lyophilized from 10 mM Sodium Acetate, pH 6.0 + 0.1 M Arginine.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.5 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthony A Portale et al.
PloS one, 10(11), e0142924-e0142924 (2015-11-21)
Fibroblast growth factor 23 (FGF23) is a potent regulator of phosphate (Pi) and vitamin D homeostasis. The transcription factor, early growth response 1 (egr-1), is a biomarker for FGF23-induced activation of the ERK1/2 signaling pathway. We have shown that ERK1/2
Richard G Boles
Handbook of Growth and Growth Monitoring in Health and Disease null
Katalin Bartos et al.
Journal of cell communication and signaling (2022-11-27)
Memo1 deletion in mice causes premature aging and an unbalanced metabolism partially resembling Fgf23 and Klotho loss-of-function animals. We report a role for Memo's redox function in renal FGF23-Klotho signaling using mice with postnatally induced Memo deficiency in the whole
Klotho converts canonical FGF receptor into a specific receptor for FGF23.
Urakawa I
Nature, 444(7120), 770-774 (2006)
Fibroblast growth factors.
Ornitz, D.M., et al.
Genome Biology, 2, 30051-300512 (2001)

Articles

Fibroblast growth factors in cell culture and various growth factors for your research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service