Skip to Content
Merck
All Photos(2)

Key Documents

SRP0364

Sigma-Aldrich

mTOR/Raptor/MLST8 human

recombinant, expressed in baculovirus infected Sf9 cells, ≥50% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥50% (SDS-PAGE)

form

aqueous solution

mol wt

137 kDa (mTOR)
150 kDa (Raptor)
37 kDa (MLST8)

packaging

pkg of 10 μg

manufacturer/tradename

Sigma-Aldrich

technique(s)

cell based assay: suitable

solubility

water: soluble

application(s)

life science and biopharma

shipped in

dry ice

storage temp.

−70°C

Gene Information

General description

This product (recombinant mTOR/Raptor/MLST8) is a complex of human mechanistic target of rapamycin (mTOR), also known as FRAP1 (GenBank Accession No. NM_004958), amino acids 1362-end with N-terminal FLAG-tag, MW = 137 kDa; human mTOR associated protein, LST8 homolog (MLST8) (GenBank Accession No. NM_022372) amino acids 2-326 (end) with N-terminal His-tag, MW = 37 kDa and human regulatory associated protein of MTOR complex 1 (Raptor) (GenBank Accession No. NM_020761) amino acids 2-1335 (end) with N-terminal His-tag, MW=150 kDa, co-expressed in a Baculovirus-infected Sf9 cell expression system.

Application

Recombinant mTOR/Raptor/MLST8 has been used in in vitro caspase reactions.

Biochem/physiol Actions

Mechanistic target of rapamycin (mTOR) modulates cell growth and division by integrating nutrient and mitogen signals. It also modulates the functions of antigen-presenting cells, like dendritic cells and activates T-cells. mTOR is also involved in innate and adaptive immunity. mTOR associated protein, LST8 homolog (MLST8) is a subunit of both the mTOR complex 1 and 2 (mTORC1 and 2). It is crucial for the activation of the mTOR kinase and is a regulator of mTOR pathways. Regulatory-associated protein of mTOR complex 1 (RAPTOR) has an important role in modulating cell growth and energy homeostasis as a scaffolding protein. It takes part in recruiting substrates of the mechanistic target of rapamycin (mTOR) to the rapamycin-sensitive mTOR complex 1 (mTORC1). RAPTOR also associates with proteins involved in the pathway and aids in phosphorylating them.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

mTOR controls cell cycle progression through its cell growth effectors S6K1 and 4E-BP1/eukaryotic translation initiation factor 4E.
Fingar DC
Molecular and Cellular Biology, 24(1), 200-216 (2004)
Immunoregulatory functions of mTOR inhibition.
Thomson AW
Nature Reviews: Immunology, 9(5), 324-337 (2009)
Belen Rubio-Viqueira et al.
Advances in experimental medicine and biology, 587, 309-327 (2006-12-14)
The mammalian target of rapamycin (mTOR) is involved in the control of cellular growth and proliferation. Abnormal activation of signaling pathways both proximal and distal to this kinase occurs frequently in human cancer suggesting that mTOR is an attractive target
mLST8 Promotes mTOR-Mediated Tumor Progression.
Kakumoto K
PLoS ONE, 10(4), e0119015-e0119015 (2015)
R Martin et al.
Cell death discovery, 2, 16024-16024 (2016-08-24)
The mammalian target of rapamycin complex 1 (mTORC1) is a highly conserved protein complex regulating key pathways in cell growth. Hyperactivation of mTORC1 is implicated in numerous cancers, thus making it a potential broad-spectrum chemotherapeutic target. Here, we characterized how

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service