Skip to Content
Merck
All Photos(1)

Key Documents

SML3706

Sigma-Aldrich

BT2

≥98% (HPLC)

Synonym(s):

3,6-Dichloro-1-benzothiophene-2-carboxylic acid, 3,6-Dichlorobenzo[b]thiophene-2-carboxylic acid, 3,6-Dichlorobenzothiophene-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H4Cl2O2S
CAS Number:
Molecular Weight:
247.10
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[s]1c2c(c(c1C(=O)O)Cl)ccc(c2)Cl

InChI

1S/C9H4Cl2O2S/c10-4-1-2-5-6(3-4)14-8(7(5)11)9(12)13/h1-3H,(H,12,13)

InChI key

AAHPIJMQJAZYTM-UHFFFAOYSA-N

Biochem/physiol Actions

BT2 is an orally active, allosteric mitochondrial branched-chain α-ketoacid dehydrogenase (BCKD) kinase (BCKDK or BDK) inhibitor (IC50 = 3.19 μM) that induces BDK dissociation from the BCKD complex (BCKDC) with superior pharmacokinetics and metabolic stability than (S)-CPP. BT2 increases cellular BCKDC activity in cultures (40 μM; MEF & murine hepatocytes) and induces BDK degradation in mice & rats in vivo, effectively upregulating tissue BCKDC activity and downregulating plasma branched-chain amino acid (BCAA) level. Typical dosing range: 40-250 μM (in cultures or cell-free), 20-40 mg/kg (ip. or po. in vivo; mice & rats) or 200 mg/kg diet.
Orally active, allosteric mitochondrial branched-chain α-ketoacid dehydrogenase complex (BCKDC) kinase (BCKDK, BDK) inhibitor in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mengping Chen et al.
Journal of the American Heart Association, 8(11), e011625-e011625 (2019-08-23)
Background Branched-chain amino acid (BCAA) catabolic defect is an emerging metabolic hallmark in failing hearts in human and animal models. The therapeutic impact of targeting BCAA catabolic flux under pathological conditions remains understudied. Methods and Results BT2 (3,6-dichlorobenzo[b]thiophene-2-carboxylic acid), a
Golam M Uddin et al.
Cardiovascular diabetology, 18(1), 86-86 (2019-07-07)
Branched chain amino acids (BCAA) can impair insulin signaling, and cardiac insulin resistance can occur in the failing heart. We, therefore, determined if cardiac BCAA accumulation occurs in patients with dilated cardiomyopathy (DCM), due to an impaired catabolism of BCAA
Danielle Murashige et al.
Cell metabolism, 34(11), 1749-1764 (2022-10-13)
Pharmacologic activation of branched-chain amino acid (BCAA) catabolism is protective in models of heart failure (HF). How protection occurs remains unclear, although a causative block in cardiac BCAA oxidation is widely assumed. Here, we use in vivo isotope infusions to show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service