Skip to Content
Merck
All Photos(1)

Key Documents

SML3687

Sigma-Aldrich

TL8-506

≥98% (HPLC)

Synonym(s):

(1E,4E)-Ethyl 2-amino-8-(3-cyanophenyl)-3H-1-benzo[b]azepine-4-carboxylate, Ethyl 2-amino-8-(3-cyanophenyl)-3H-1-benzazepine-4-carboxylate, TL 8-506, TLR 8-506

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17N3O2
CAS Number:
Molecular Weight:
331.37
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

TL8-506 is a motolimod (VTX-2337) analog and a benzoazepine class toll-like receptor 8 (TLR8)-selective agonist. TL8-506 is commonly employed in the concentration range from 0.1 μg/mL to 1 μg/mL for stimulating cell surface TLR8 in cultures and is reported to be a stronger TLR8 ligand than the imidazoquinoline (IMDQ) derivatives (IQDs) resiquimod (R848) and CL075 (3M002).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer Nelson et al.
Science advances, 6(26), eaaz6893-eaaz6893 (2020-07-09)
Messenger RNA (mRNA) represents an attractive therapeutic modality for potentially a wide range of clinical indications but requires uridine chemistry modification and/or tuning of the production process to prevent activation of cellular innate immune sensors and a concomitant reduction in
Bin Tang et al.
Parasitology research, 119(8), 2505-2510 (2020-06-15)
Human trichinellosis is acquired by eating raw or undercooked meats carrying muscle larvae of Trichinella spp. Toll-like receptors (TLRs) are essential components of the innate immune system. However, little is known about the potential application of TLR agonists for immunotherapy
Jahnavi Aluri et al.
Blood, 137(18), 2450-2462 (2021-01-30)
Inborn errors of immunity (IEI) are a genetically heterogeneous group of disorders with a broad clinical spectrum. Identification of molecular and functional bases of these disorders is important for diagnosis, treatment, and an understanding of the human immune response. We
Kamil Krawczynski et al.
Journal of cell science, 134(5) (2020-10-24)
The function of microRNAs (miRNAs) can be cell autonomous or communicated to other cell types and has been implicated in diverse biological processes. We previously demonstrated that miR-517a-3p (miR-517a), a highly expressed member of the chromosome 19 miRNA cluster (C19MC)
Mahesh Chandra Patra et al.
Cells, 9(7) (2020-07-15)
Toll-like receptors (TLRs) play a fundamental role in the inflammatory response against invading pathogens. However, the dysregulation of TLR-signaling pathways is implicated in several autoimmune/inflammatory diseases. Here, we show that a novel small molecule TLR-inhibitor (TAC5) and its derivatives TAC5-a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service