Skip to Content
Merck
All Photos(1)

Key Documents

SML3602

Sigma-Aldrich

Pipobroman

≥98% (HPLC)

Synonym(s):

1,​1′-​(1,​4-​Piperazinediyl)​bis[3-​bromo-1-​propanone, 1,​4-​bis(3-​Bromo-​1-​oxopropyl)​-piperazine, N,N-Bis-(3-bromopropionyl)-piperazine, NSC-​25154

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16Br2N2O2
CAS Number:
Molecular Weight:
356.05
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

BrCCC(=O)N1CCN(CC1)C(=O)CCBr

InChI

1S/C10H16Br2N2O2/c11-3-1-9(15)13-5-7-14(8-6-13)10(16)2-4-12/h1-8H2

InChI key

NJBFOOCLYDNZJN-UHFFFAOYSA-N

Biochem/physiol Actions

Anti-cancer drug, cytostatic alkylating agent
Pipobroman is an alkylating agent that is used as anti-cancer drug. It also has clinical activity in polycythemia vera and thrombocythemia.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean-Jacques Kiladjian et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 29(29), 3907-3913 (2011-09-14)
The overall impact of hydroxyurea (HU) or pipobroman treatments on the long-term outcome of patients with polycythemia vera (PV) has not been assessed in randomized studies. We report final analyses from the French Polycythemia Study Group (FPSG) study, which randomly
Giovanni Fazio et al.
Journal of thrombosis and thrombolysis, 35(2), 282-285 (2012-07-31)
Polycythemia vera (PV) is a chronic myeloproliferative disorder characterized by an abnormal increase in red blood cells. The involvement of the heart during the course of the illness represents a common cause of morbidity and it is linked to an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service