Skip to Content
Merck

F0430

Fenobam

≥98% (HPLC), solid

Synonym(s):

N-(3-Chlorophenyl)-N′-(4,5-dihydro-1-methyl-4-oxo-1H-imidazole-2-yl)urea

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H11ClN4O2
CAS Number:
Molecular Weight:
266.68
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: >20 mg/mL

originator

Johnson & Johnson

storage temp.

2-8°C

SMILES string

CN1CC(=O)N=C1NC(=O)Nc2cccc(Cl)c2

InChI

1S/C11H11ClN4O2/c1-16-6-9(17)14-10(16)15-11(18)13-8-4-2-3-7(12)5-8/h2-5H,6H2,1H3,(H2,13,14,15,17,18)

InChI key

DWPQODZAOSWNHB-UHFFFAOYSA-N

Biochem/physiol Actions

Fenobam is a potent, selective, noncompetitive glutamate mGluR5 receptor antagonist. Fenobam displays inverse agonist properties; blocks mGluR5 constitutive activity in vitro (IC50 = 87 nM, slightly weaker than MPEP). Fenobam acts at an allosteric modulatory site shared with MPEP and binds the mGlu5 receptor with Kd values of 54 and 31 nM for rat and human receptors, respectively. Fenobam belongs to a structurally different class than MPEP; devoid of GABAergic activity and thus typical benzodiazepine-like side effects; displays anxiolytic activity.
Potent, selective, noncompetitive metabotropic glutamate receptor antagonist (mGluR5). Displays anxiolytic activity.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Glutamate Receptors (G Protein Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.


Still not finding the right product?

Explore all of our products under Fenobam


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library