Skip to Content
Merck
All Photos(1)

Key Documents

5.33916

Sigma-Aldrich

KIFC1 Inhibitor, AZ82

Synonym(s):

KIFC1 Inhibitor, AZ82, 5-Methyl-N-(( R)-1-oxo-1-(( R)-pyrrolidin-3-ylamino)-3-(6-(3-(trifluoromethoxy)phenyl)pyridin-3-yl)propan-2-yl)-4-propylthiophene-2-carboxamide, Tctex7 Inhibitor, Knsl2 Inhibitor, MT/KIFC1 Inhibitor, Bm4137 Inhibitor, HSET Inhibitor, Kinesi, 5-Methyl-N-((R)-1-oxo-1-((R)-pyrrolidin-3-ylamino)-3-(6-(3-(trifluoromethoxy)phenyl)pyridin-3-yl)propan-2-yl)-4-propylthiophene-2-carboxamide, Tctex7 Inhibitor, Knsl2 Inhibitor, MT/KIFC1 Inhibitor, Bm4137 Inhibitor, HSET Inhibitor, Kinesin F

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H31F3N4O3S
CAS Number:
Molecular Weight:
560.63
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥96% (HPLC)

Quality Level

form

film

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable pyridinylpropanylthiophene-carboxamide compound that acts as a potent, reversible, and ATP-competitive (Ki = 43 nM) and microtubule (MT) non-competitive inhibitor of MT-stimulated ATPase activity of KIFC1 (IC50 = 300 nM). However, it does not affect the basal ATPase activity even at high concentrations (~100 µM). Binds to the MT/KIFC1 complex (Kd of 690 nM) with high-affinity and blocks ATP binding and release of ADP. Does not directly interact with KIFC protein that is not associated with MT. Exhibits high degree of selectivity against a panel of nine kinesin motor proteins at a concentration (~ 5 µM) where KIFC1 activity is reduced by 95%. Causes mitotic delay, multipolar spindle formation and centrosome declustering in centrosome-amplified BT-549 cancer cells without affecting normal cells. Also shown to prevent Eg5 inhibition induced monopolar spindles and restores bipolar mitotic spindle formation in HeLa cells at 400 nM.
A cell-permeable pyridinylpropanylthiophene-carboxamide compound that exhibits selective affinity toward MT/microtubule-bound KIFC1 motor domain (Q305-K673; Kd = 690 nM), but not to uncomplexed KIFC1 or MT, and selectively inhibits MT-induced, but not basal, KIFC1 motor domain ATPase activity (IC50 = 300 nM; [KIFC1] = 30 nM, [MT] = 100 nM, [ATP] = 15 µM) in an ATP-competitive (Ki = 43 nM; [KIFC1] = 125 nM, [MT] = 1.5 µM) and MT-noncompetitive manner, effectively preventing ADP release, while displaying little potency against 9 other kinesin motor proteins (<15% or no inhibition at 5 µM against human CENP-E, Chromokinesin/KIF4A, Eg5, KIFC3, KIF3C, KInesin Heavy Chain/KIF5B, MCAK/KIF2C, MKLP1/KIF23, and A. nidulans BimC/KIF8). KIFC1 knockdown by siRNA or functional inhibition by AZ82 (0.4 to 1.2 µM) is shown to result in mitotic delay and multipolar spindle formation among BT549 population with extra centrosomes due to failure of chromosomes clustering. In cells with normal/two centrosomes, Eg5 inhibition causes monopolar spindles formation, while AZ82 co-treatment (400 nM) offsets the effect of Eg5 inhibition (5 nM AZD4877) and restores bipolar spindles phenotype in mitotic HeLa cells. Off-target cytotoxicity is reported when used at concentrations above 4 µM.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MT/KIFC1
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Reference: Wu, J., et al. 2013. ACS Chem. Biol.8, 2201.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service