Skip to Content
Merck
All Photos(4)

Documents

M9511

Sigma-Aldrich

Minocycline hydrochloride

powder, antibiotic

Synonym(s):

Minomycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27N3O7 · HCl
CAS Number:
Molecular Weight:
493.94
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Minocycline hydrochloride, powder

form

powder

Quality Level

antibiotic activity spectrum

mycobacteria

Mode of action

protein synthesis | interferes

originator

Teva

storage temp.

2-8°C

SMILES string

Cl.CN(C)[C@H]1[C@@H]2C[C@@H]3Cc4c(ccc(O)c4C(=O)C3=C(O)[C@]2(O)C(=O)C(C(N)=O)=C1O)N(C)C

InChI

1S/C23H27N3O7.ClH/c1-25(2)12-5-6-13(27)15-10(12)7-9-8-11-17(26(3)4)19(29)16(22(24)32)21(31)23(11,33)20(30)14(9)18(15)28;/h5-6,9,11,17,27,29-30,33H,7-8H2,1-4H3,(H2,24,32);1H/t9-,11-,17-,23-;/m0./s1

InChI key

GLMUAFMGXXHGLU-VQAITOIOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: tetracycline
Minocycline, a semi-synthetic second-generation tetracycline, is classified within the tetracycline class of antibiotics and is utilized for the management and treatment of both infectious and non-infectious conditions. Apart from its efficacy against gram-positive and gram-negative bacteria, minocycline also exhibits antioxidant, anti-apoptotic, immunomodulatory, and neuroprotective properties. Tetracyclines, including minocycline, function by binding to the 30S ribosomal subunit, thereby preventing charged tRNA from delivering amino acids to elongate the protein chain and form cellular protein. This disruption results in a bacteriostatic effect on the prokaryotic cell, leading to the loss of its ability to grow or replicate. Being lipid-soluble compounds, tetracyclines can traverse hydrophobic barriers such as biological membranes. Among tetracyclines, minocycline demonstrates higher lipophilicity than doxycycline, leading to increased concentrations in the central nervous system (CNS) and the skin.

Application

Minocycline hydrochloride has been used:
  • to prepare nanoliposomes to check its effect on macrophages
  • for the inhibition of neuroinflammation and neuropathic pain
  • to treat glioma in murine GL261 glioma cells

Biochem/physiol Actions

Minocycline is a broad spectrum antibiotic with bacteriostatic function. Minocycline has anti-inflammatory properties. Minocycline inhibits lipopolysaccharide mediated inflammatory cytokine tumour necrosis factor (TNF-α) secretion by macrophages. Minocycline inhibits macrophage proliferation in a dose dependent manner. Minocycline inhibits neuroinflammation in pre-plaque of Alzheimer′s disease-like amyloid pathology through inhibition of key inflammatory enzymes like inducible nitric oxide synthase (iNOS), matrix metalloproteinase 9 (MMP-9) and 5-lipoxygenase. Minocycline inhibits endothelial cell proliferation and angiogenesis. Minocycline exhibits anti-tumor activity in glioma by inhibiting membrane type 1 matrix metalloproteinase (MT1-MMP). Minocycline increases cognition and neuronal differentiation. zMinocycline effectively reduces neuropathic pain by increasing the functions of nociceptin/orphanin FQ.

Features and Benefits

This compound was developed by Teva. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Jin-Shing Chen et al.
Lancet (London, England), 381(9874), 1277-1282 (2013-03-16)
Simple aspiration and drainage is a standard initial treatment for primary spontaneous pneumothorax, but the rate of pneumothorax recurrence is substantial. We investigated whether additional minocycline pleurodesis after simple aspiration and drainage reduces the rate of recurrence. In our open-label
Minocycline enhances the effectiveness of nociceptin/orphanin FQ during neuropathic pain
Popiolek-Barczyk K, et al.
BioMed Research International, 2014 (2014)
Minocycline corrects early, pre-plaque neuroinflammation and inhibits BACE-1 in a transgenic model of Alzheimer's disease-like amyloid pathology
Ferretti MT, et al.
Journal of Neuroinflammation, 9(1), 62-62 (2012)
Minocycline
Nazarian S and Akhondi H
StatPearls [Internet] (2023)
Xuejiao Wang et al.
Journal of neuroimmunology, 334, 576994-576994 (2019-06-18)
Central nervous system (CNS) involvement is commonly seen in the patients with system lupus erythematosus (SLE). Mechanisms underlying CNS damage in SLE remain largely unknown. Accumulating evidence suggest that activation of microglia in CNS plays an important role in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service