Skip to Content
Merck
All Photos(1)

Key Documents

SML2836

Sigma-Aldrich

Lixisenatide

≥95% (HPLC)

Synonym(s):

(Des-Pro38)-Exendin-4-(Lys)6 amide, AVE0010, HGEGTFTSDLSKQMEEEAVRLFIEWLKNGGPSSGAPPSKKKKKK-NH2, His-Gly-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Ser-Lys-Lys-Lys-Lys-Lys-Lys-NH2, ZP10, ZP10A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C215H347N61O65S
CAS Number:
Molecular Weight:
4858.49
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

lyophilized powder

color

white to beige

storage temp.

−20°C

Biochem/physiol Actions

Exendin-4-derived glucagon-like peptide-1 receptor (GLP-1R) agonist with in vivo therapeutic efficacy in animal models of type 2 diabetes.
Lixisenatide (AVE0010, ZP10A) is a C-terminal amidated synthetic glucagon-like peptide-1 receptor (GLP-1R) agonist peptide whose sequence corresponds to Pro38 deleted exendin-4 with a C-terminal extension by six Lys residues. Lixisenatide exhibits 4-times higher human GLP-1R affinity than GLP-1(7-36) amide and dispalys in vivo therapeutic efficacy in murine and rat models of type 2 diabetes, as well as rat models of dox-induced renal fibrosis, global cerebral I/R injury, abdominal aortic aneurysm (AAA) and Aβ25-35 toxicity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qini Zhao et al.
Artificial cells, nanomedicine, and biotechnology, 47(1), 2325-2332 (2019-06-09)
Increased free fatty acids (FFA) are one of the risk factors for type 2 diabetes. FFA also contribute to endothelial dysfunction in both the prediabetes and diabetes conditions. Therefore, FFA are an important link between diabetes and endothelial dysfunction. In
Preclinical pharmacology of the new GLP-1 receptor agonist AVE0010.
U Werner
Annales d'endocrinologie, 69(2), 164-165 (2008-04-18)
Julie Charpentier et al.
American journal of physiology. Gastrointestinal and liver physiology, 315(5), G671-G684 (2018-08-03)
Endogenous glucagon-like peptide-1 (GLP-1) regulates glucose-induced insulin secretion through both direct β-cell-dependent and indirect gut-brain axis-dependent pathways. However, little is known about the mode of action of the GLP-1 receptor agonist lixisenatide. We studied the effects of lixisenatide (intraperitoneal injection)
Jie Yu et al.
Surgery today, 46(9), 1099-1107 (2015-12-15)
To demonstrate the protective effect of glucagon-like peptide 1 (GLP-1) signaling on the cardiovascular system, we conducted this study to show that the GLP-1 receptor analog (lixisenatide) could inhibit abdominal aortic aneurysm (AAA) development in rats. Lixisenatide was injected subcutaneously
Zhen Zhao et al.
Biochemical and biophysical research communications, 508(4), 1120-1125 (2018-12-17)
Mitochondrial dysregulation has been associated with vascular endothelial dysfunction and pathophysiological development of cardiovascular diseases. Lixisenatide is a drug approved by the US Food and Drug Administration for the treatment of type 2 diabetes (T2D). Little information regarding the effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service