Skip to Content
Merck
All Photos(1)

Key Documents

G3265

Sigma-Aldrich

Glucagon-Like Peptide I human

≥97% (HPLC)

Synonym(s):

GLP-1, Preproglucagon 72-108

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C186H275N51O59
CAS Number:
Molecular Weight:
4169.48
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

InChI

1S/C186H275N51O59/c1-17-94(10)149(182(294)209-98(14)155(267)220-128(73-105-78-199-111-42-28-27-41-109(105)111)171(283)223-123(68-91(4)5)173(285)234-147(92(6)7)180(292)219-113(43-29-31-63-187)158(270)200-81-136(245)210-112(45-33-65-197-185(191)192)157(269)203-84-146(262)263)236-174(286)126(70-102-37-23-19-24-38-102)225-165(277)120(55-61-142(254)255)216-162(274)114(44-30-32-64-188)212-153(265)96(12)206-152(264)95(11)207-161(273)118(51-57-135(190)244)211-137(246)82-201-160(272)117(53-59-140(250)251)215-168(280)122(67-90(2)3)222-170(282)125(72-104-47-49-108(243)50-48-104)226-177(289)132(85-238)230-179(291)134(87-240)231-181(293)148(93(8)9)235-176(288)131(77-145(260)261)228-178(290)133(86-239)232-184(296)151(100(16)242)237-175(287)127(71-103-39-25-20-26-40-103)229-183(295)150(99(15)241)233-138(247)83-202-159(271)116(52-58-139(248)249)213-154(266)97(13)208-167(279)129(75-107-80-196-89-205-107)227-163(275)115(46-34-66-198-186(193)194)214-164(276)119(54-60-141(252)253)217-169(281)124(69-101-35-21-18-22-36-101)224-166(278)121(56-62-143(256)257)218-172(284)130(76-144(258)259)221-156(268)110(189)74-106-79-195-88-204-106/h18-28,35-42,47-50,78-80,88-100,110,112-134,147-151,199,238-243H,17,29-34,43-46,51-77,81-87,187-189H2,1-16H3,(H2,190,244)(H,195,204)(H,196,205)(H,200,270)(H,201,272)(H,202,271)(H,203,269)(H,206,264)(H,207,273)(H,208,279)(H,209,294)(H,210,245)(H,211,246)(H,212,265)(H,213,266)(H,214,276)(H,215,280)(H,216,274)(H,217,281)(H,218,284)(H,219,292)(H,220,267)(H,221,268)(H,222,282)(H,223,283)(H,224,278)(H,225,277)(H,226,289)(H,227,275)(H,228,290)(H,229,295)(H,230,291)(H,231,293)(H,232,296)(H,233,247)(H,234,285)(H,235,288)(H,236,286)(H,237,287)(H,248,249)(H,250,251)(H,252,253)(H,254,255)(H,256,257)(H,258,259)(H,260,261)(H,262,263)(H4,191,192,197)(H4,193,194,198)/t94-,95-,96-,97-,98-,99+,100+,110-,112-,113-,114-,115-,116-,117-,118-,119-,120-,121-,122-,123-,124-,125-,126-,127-,128-,129-,130-,131-,132-,133-,134-,147-,148-,149-,150-,151-/m0/s1

InChI key

UKVFVQPAANCXIL-FJVFSOETSA-N

Gene Information

human ... GCG(2641)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

His-Asp-Glu-Phe-Glu-Arg-His-Ala-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser-Tyr-Leu-Glu-Gly-Gln-Ala-Ala-Lys-Glu-Phe-Ile-Ala-Trp-Leu-Val-Lys-Gly-Arg-Gly

General description

Glucagon-Like Peptide I (GLP1) is an incretin hormone, which functions as ligand for GLP1 receptors (GLP1R). These receptors are expressed in renal and cardiovascular system, such as smooth muscle and endothelial cells.

Application

Glucagon-Like Peptide I (GLP1) has been used as an antigen in excess amounts for the incubation of primary antibodies, post-immunocytochemistry with positive results of paraffin-embedded pancreatic tissues of Rana temporaria.
Glucagon-Like Peptide I human has been used as a component of CMRL1066 medium for culturing human pancreatic non-endocrine cells. It has also been used for preabsoption of antibodies prior to immunoassay

Biochem/physiol Actions

Glucagon-Like Peptide I (GLP1) controls the metabolism of glucose, by inducing the secretion of insulin and suppressing that of glucagon. Studies in Indonesian population show that reduced levels of this protein are linked with increased susceptibility to type 2 diabetes mellitus. Plasma levels of this hormone also show significant association with systolic and diastolic blood pressure levels.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C Etayo et al.
General and comparative endocrinology, 117(3), 366-380 (2000-04-15)
To characterize the endocrine cell types of the pancreas of Rana temporaria, conventional staining, silver impregnation, and immunocytochemical methods for light and electron microscopy have been applied to paraffin, thin and semithin sections, many of them serial pairs. Quantitative data
S Sarkar et al.
General and comparative endocrinology, 121(1), 23-31 (2001-02-13)
The organization of glucagon-like immunoreactivity (GLI) in the olfactory system, forebrain, and pituitary was investigated in the teleost Clarias batrachus. Weak to moderate GLI was seen in some olfactory receptor neurons and basal cells of the olfactory epithelium. Intense GLI
Exon duplication and divergence in the human preproglucagon gene.
G I Bell et al.
Nature, 304(5924), 368-371 (1983-07-03)
Agus Lastya et al.
BMC research notes, 7, 849-849 (2014-11-28)
Glucagon like peptide-1 (GLP-1), an incretin hormone, regulates glucose metabolism by inducing insulin secretion and suppressing glucagon secretion. The aim of the study is to assess the levels of fasting and post-prandial GLP-1 and their risk for T2DM. A case
Philipp Krisai et al.
Hypertension (Dallas, Tex. : 1979), 65(2), 306-312 (2014-12-03)
Hypertension and diabetes mellitus are highly correlated, but the underlying mechanisms are only partly understood. Therefore, the aim of our study was to investigate the relationships between plasma levels of glucagon-like peptide-1, a key factor in the regulation of glucose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service