Skip to Content
Merck
All Photos(1)

Key Documents

SML2647

Sigma-Aldrich

CA3

≥97% (HPLC)

Synonym(s):

1,1′-[[9-(Hydroxyimino)-9H-fluorene-2,7-diyl]bis(sulfonyl)]bis-piperidine, 2,7-bis(1-Piperidinylsulfonyl)-9H-fluoren-9-one oxime, CIL56

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27N3O5S2
CAS Number:
Molecular Weight:
489.61
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[S](=O)(=O)(N5CCCCC5)c1cc2c(cc1)c3c(cc(cc3)[S](=O)(=O)N4CCCCC4)C2N=O

InChI

1S/C23H27N3O5S2/c27-24-23-21-15-17(32(28,29)25-11-3-1-4-12-25)7-9-19(21)20-10-8-18(16-22(20)23)33(30,31)26-13-5-2-6-14-26/h7-10,15-16,23H,1-6,11-14H2

InChI key

YLRXSLQNJCJWGZ-UHFFFAOYSA-N

Biochem/physiol Actions

CA3 is a potent and selective inhibitor of YAP1 (Hippo co-activator Yes-associated protein 1) that inhibits YAP1 expression and transcriptional activity in esophageal adenocarcinoma (EAC) cell lines. CA3 potently inhibits growth, induces apoptosis in radiation resistant CSC (cancer stem cell) enriched EAC cells. It inhibits tumor growth of in xenograft model with no apparent toxicity.
potent and selective inhibitor of YAP1 that inhibits YAP1 expression and transcriptional activity in EAC cell lines

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shumei Song et al.
Molecular cancer therapeutics, 17(2), 443-454 (2017-11-24)
Mounting evidence suggests that the Hippo coactivator Yes-associated protein 1 (YAP1) is a major mediator of cancer stem cell (CSC) properties, tumor progression, and therapy resistance as well as often a terminal node of many oncogenic pathways. Thus, targeting YAP1
Xiao Qin et al.
Cell, 186(25), 5554-5568 (2023-12-09)
Cancer cells are regulated by oncogenic mutations and microenvironmental signals, yet these processes are often studied separately. To functionally map how cell-intrinsic and cell-extrinsic cues co-regulate cell fate, we performed a systematic single-cell analysis of 1,107 colonic organoid cultures regulated
Siyuan Hao et al.
iScience, 27(6), 109927-109927 (2024-05-24)
YAP/TEAD signaling is essential for organismal development, cell proliferation, and cancer progression. As a transcriptional coactivator, how YAP activates its downstream target genes is incompletely understood. YAP forms biomolecular condensates in response to hyperosmotic stress, concentrating transcription-related factors to activate
Sina Stern et al.
Neuron, 109(21), 3436-3455 (2021-09-12)
An inhibitory extracellular milieu and neuron-intrinsic processes prevent axons from regenerating in the adult central nervous system (CNS). Here we show how the two aspects are interwoven. Genetic loss-of-function experiments determine that the small GTPase RhoA relays extracellular inhibitory signals

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service