Skip to Content
Merck
All Photos(1)

Key Documents

SML2557

Sigma-Aldrich

Aminopyridine 2

≥98% (HPLC)

Synonym(s):

1-((6-Aminopyridin-3-yl)methyl)-3-(4-bromophenyl)urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13BrN4O
CAS Number:
Molecular Weight:
321.17
UNSPSC Code:
12352200
NACRES:
NA.77

Pricing and availability is not currently available. Contact Technical Service

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=CC=C(CNC(NC2=CC=C(Br)C=C2)=O)C=N1

InChI

1S/C13H13BrN4O/c14-10-2-4-11(5-3-10)18-13(19)17-8-9-1-6-12(15)16-7-9/h1-7H,8H2,(H2,15,16)(H2,17,18,19)

Biochem/physiol Actions

Aminopyridine 2 (1-((6-aminopyridin-3-yl)methyl)-3-(4-bromophenyl)urea) is an orally available, potent and selective irreversible inhibitor of myeloperoxidase (MPO) that blocks vasomotor dysfunction in rat aortic rings. Aminopyridine 2 inhibits myeloperoxidase in a mouse model of peritonitis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service