Skip to Content
Merck
All Photos(1)

Key Documents

SML2394

Sigma-Aldrich

AT9283

≥98% (HPLC)

Synonym(s):

1-Cyclopropyl-3-[3-(5-morpholin-4-ylmethyl-1H-benzoimidazol-2-yl)-1H-pyrazol-4-yl]-urea, AT 9283, AT-9283, N-Cyclopropyl-N′-[3-[6-(4-morpholinylmethyl)-1H-benzimidazol-2-yl]-1H-pyrazol-4-yl]-urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23N7O2
CAS Number:
Molecular Weight:
381.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[nH]1nc(c(c1)NC(=O)NC5CC5)c2[nH]c3c(n2)ccc(c3)CN4CCOCC4

InChI

1S/C19H23N7O2/c27-19(21-13-2-3-13)24-16-10-20-25-17(16)18-22-14-4-1-12(9-15(14)23-18)11-26-5-7-28-8-6-26/h1,4,9-10,13H,2-3,5-8,11H2,(H,20,25)(H,22,23)(H2,21,24,27)

InChI key

LOLPPWBBNUVNQZ-UHFFFAOYSA-N

Biochem/physiol Actions

A multitargeted kinase inhibitor with in vivo anticancer efficacy and potent activity against Abl, aurora kinase A/B and JAK2/3.
AT9283 has also been shown to accelerate growth arrest in various xenograft models.
AT9283 is an ATP site-targeting pyrazole-benzimidazole-based molecule with inhibitory potency against multiple kinases, most notably Aurora A/B (IC50 ≤3.0 nM), JAK2/3 (IC50 = 1.2/1.1 nM), Abl T315I (IC50 = 4 nM), GSK3β, FGFR2, VEGFR3 (Flt4), Mer, Ret, Rsk2/3, Tyk2, Yes (IC50 = 1-10 nM). AT9283 also inhibits 72 other kinases/mutations at a reduced potency (IC50 = 10-30 nM against 14, 30-100 nM against 21, 100-300 against 37 targets), while displaying an IC50 >300 nM toward 144 other kinase targets. AT9283 inhibits the growth/survival of multiple solid tumor cell lines in vitro (IC50 = 7.7-20 nM) and shows anti-cancer efficacy in mice via i.p. injection in vivo (67% and 76%HCT116 tumor growth inhibition on day 16, respectively, with 15 and 20 mg/kg b.i.d., two-day on and two-day off; >95% inhibition of Ba/F3 ETV6-JAK2 leukemia proliferation on day 12 with 10 mg/kg b.i.d. on days 2–5 and 8–12).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanne H Olesen et al.
ChemMedChem, 11(11), 1137-1144 (2016-05-03)
Increasing evidence suggests key roles for members of the mammalian Sterile20-like (MST) family of kinases in many aspects of biology. MST3 is a member of the STRIPAK complex, the deregulation of which has recently been associated with cancer cell migration
Mark A Dawson et al.
British journal of haematology, 150(1), 46-57 (2010-05-29)
Constitutive activation of Janus kinase (Jak) 2 is the most prevalent pathogenic event observed in the myeloproliferative disorders (MPD), suggesting that inhibitors of Jak2 may prove valuable in their management. Inhibition of the Aurora kinases has also proven to be
Fiona J Sorrell et al.
Structure (London, England : 1993), 24(3), 401-411 (2016-02-09)
The highly diverse Numb-associated kinase (NAK) family has been linked to broad cellular functions including receptor-mediated endocytosis, Notch pathway modulation, osteoblast differentiation, and dendrite morphogenesis. Consequently, NAK kinases play a key role in a diverse range of diseases from Parkinson's
Targeting Protein Kinases for Cancer Therapy (2011)
Steven Howard et al.
Journal of medicinal chemistry, 52(2), 379-388 (2009-01-16)
Here, we describe the identification of a clinical candidate via structure-based optimization of a ligand efficient pyrazole-benzimidazole fragment. Aurora kinases play a key role in the regulation of mitosis and in recent years have become attractive targets for the treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service