Skip to Content
Merck
All Photos(1)

Key Documents

SML1406

Sigma-Aldrich

Brassinazole

≥98% (HPLC), powder, brassinosteroid biosynthesis inhibitor

Synonym(s):

β-[(4-chlorophenyl)methyl]-α-methyl-α-phenyl-1H-1,2,4-triazole-1-ethanol, (αRS)-rel-β-[(4-chlorophenyl)methyl]-α-methyl-α-phenyl-1H-1,2,4-triazole-1-ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18ClN3O
CAS Number:
Molecular Weight:
327.81
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Brassinazole, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

ClC1=CC=C(C[C@@H](N2N=CN=C2)[C@@](C)(O)C3=CC=CC=C3)C=C1

InChI

1S/C18H18ClN3O/c1-18(23,15-5-3-2-4-6-15)17(22-13-20-12-21-22)11-14-7-9-16(19)10-8-14/h2-10,12-13,17,23H,11H2,1H3/t17-,18+/m1/s1

InChI key

YULDTPKHZNKFEY-MSOLQXFVSA-N

Application

Brassinazole has been used in phenotype analyses to detect morphological changes in G-protein β subunit agb1 mutants.[1]

Biochem/physiol Actions

Brassinazole induces dwarfism and morphological changes in Arabidopsis, which results in similarity with brassinosteroid deficient mutants. These changes were neutralized by brassinolide treatment, indicating that brassinazole leads to brassinolide deficiency in Arabidopsis.[2]
Brassinazole is an inhibitor of the biosynthesis of brassinosteroids, steroid hormones essential for plant growth and development.
Brassinazole is an inhibitor of the biosynthesis of brassinosteroids, steroid hormones essential for plant growth and development. It has been used to study brassinosteroids function.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Asami et al.
Plant physiology, 123(1), 93-100 (2000-05-12)
Screening for brassinosteroid (BR) biosynthesis inhibitors was performed to find chemicals that induce dwarfism in Arabidopsis, mutants that resembled BR biosynthesis mutants that can be rescued by BR. Through this screening experiment, the compound brassinazole was selected as the most
Lijiang Hou et al.
Plant & cell physiology, 60(8), 1761-1777 (2019-05-18)
Brassinosteroid (BR) plays an important role in plant development and biotic and abiotic stress tolerance, but its specific function remains largely unknown in wheat (Triticum aestivum L.), preventing its utilization in this important crop. In this study, the function of
Zhongtao Jia et al.
Plant physiology, 183(3), 998-1010 (2020-05-14)
Root developmental plasticity enables plants to adapt to limiting or fluctuating nutrient conditions in the soil. When grown under nitrogen (N) deficiency, plants develop a more exploratory root system by increasing primary and lateral root length. However, mechanisms underlying this
Arabidopsis G-Protein β Subunit AGB1 Interacts with BES1 to Regulate Brassinosteroid Signaling and Cell Elongation.
Zhang T, et al.
Frontiers in Plant Science, 8, 2225-2225 (2018)
Xuejiao Cheng et al.
The Plant cell, 32(4), 923-934 (2020-02-16)
Six subspecies of hexaploid wheat (Triticumaestivum) have been identified, but the origin of Indian dwarf wheat (Triticum sphaerococcum), the only subspecies with round grains, is currently unknown. Here, we isolated the grain-shape gene Tasg-D1 in Tsphaerococcum via positional cloning. Tasg-D1

Questions

1–2 of 2 Questions  
  1. What is the function, applicable concentration, and preparation method for this product?

    1 answer
    1. Brassinazole is a selective triazole-type brassinosteroid (BR) biosynthesis inhibitor used for regulating plant growth and development. The working concentration of the product would have to be determined by the end user, and the minimum purity specification is 97.5%. The compound is soluble in DMSO at 5mg/mL. For more information on the topic, it may be helpful to review the relevant publications.

      Helpful?

  2. What is the function, applicable concentration, and preparation method for this product?

    1 answer
    1. Brassinazole is a selective triazole-type brassinosteroid (BR) biosynthesis inhibitor. Brassinazole is used for regulating plant growth and development. It may be helpful to review the following publications below to obtain more information on the topic: https://www.sigmaaldrich.com/search/sml1406focus=papers&page=1&perpage=30&sort=relevance&term=sml1406&type=citation_search The working concentration of the product would have to be determined by the end user. The minimum purity specification is 97.5%. The compound is soluble in DMSO at 5mg/mL.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service