Skip to Content
Merck
All Photos(1)

Key Documents

SML1355

Sigma-Aldrich

PHTPP

≥98% (HPLC)

Synonym(s):

2-Phenyl-3-(4-hydroxyphenyl)-5,7-bis(trifluoromethyl)-pyrazolo[1,5-a]pyrimidine, 4-[2-Phenyl-5,7-bis(trifluoromethyl)pyrazolo[1,5-a]-pyrimidin-3-yl]phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H11F6N3O
CAS Number:
Molecular Weight:
423.31
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

OC(C=C1)=CC=C1C(C(C2=CC=CC=C2)=N3)=C4N3C(C(F)(F)F)=CC(C(F)(F)F)=N4

InChI

1S/C20H11F6N3O/c21-19(22,23)14-10-15(20(24,25)26)29-18(27-14)16(11-6-8-13(30)9-7-11)17(28-29)12-4-2-1-3-5-12/h1-10,30H

InChI key

AEZPAUSGTAHLOQ-UHFFFAOYSA-N

Biochem/physiol Actions

PHTPP is a selective estrogen receptor β (ERβ) full antagonist with 36-fold selectivity for ERβ over ERα. PHTPP has been used to distinguish the various activites of the two estrogen receptors. Estrogen can have opposite effects in some tumors expressing ERα compared to Erβ, with Erα enhancing and Erβ suppressing tumor cell growth. PHTPP, a selective ERβ antagonist, significantly enhanced cell growth in ovarian cancer cell lines SKOV3 and OV2008 that express both receptors.
PHTPP is a selective estrogen receptor β (ERβ) full antagonist.
PHTPP is also termed as 4-[2–phenyl-5,7–bis (trifluoromethyl) pyrazolo [1,5-a]-pyrimidin-3-yl] phenol (PHTPP). It blocks hypoglycemic augmentation of norepinephrine (NE) in the hypothalamic arcuate (ARH).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hindbrain estrogen receptor-beta antagonism normalizes reproductive and counter-regulatory hormone secretion in hypoglycemic steroid-primed ovariectomized female rats.
Briski KP and Shrestha PK
Neuroscience, 331, 62-71 (2016)
Masayo Hirao-Suzuki et al.
Molecular pharmacology, 95(3), 260-268 (2018-12-16)
Bisphenol A (BPA), recognized as an endocrine disruptor, is thought to exert its activity through a mechanism involving the activation of estrogen receptors (ERs) α/β However, a major problem is that very high concentrations of BPA are required (i.e., those
Cheng Xu et al.
Environmental science and pollution research international, 24(15), 13414-13423 (2017-04-08)
Perfluorooctane sulfonate (PFOS), an artificial fluorosurfactant and global contaminant, is used widely in various consumer products. In this study, we investigated the function of estrogen receptor β (ERβ) in PFOS-induced bile acid and cholesterol metabolism disorders and gut microbiome using
Chin-Hee Song et al.
PloS one, 14(8), e0221650-e0221650 (2019-08-24)
Several reports indicate crosstalk between the transcription factor nuclear factor erythroid 2-related factor 2 (Nrf2) and estrogen, which has a protective effect in colorectal cancer (CRC). The aim of this study was to investigate the role of Nrf2 signaling in
Qinghai Meng et al.
Journal of advanced research, 28, 149-164 (2020-12-29)
Excessive inflammation and the pyroptosis of vascular endothelial cells caused by estrogen deficiency is one cause of atherosclerosis in post-menopausal women. Because autophagy is highly regulated by estrogen, we hypothesized that estrogen can reduce vascular endothelial cell pyroptosis through estrogen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service