Skip to Content
Merck
All Photos(1)

Key Documents

I4409

Sigma-Aldrich

Fulvestrant

>98% (HPLC)

Synonym(s):

(7α,17β)-7-[9-[(4,4,5,5,5-Pentafluoropentyl)sulfinyl]nonyl]estra-1,3,5(10)-triene-3,17-diol, Faslodex, ICI 182,780, ZD 182780, ZD 9238, ZM 182780

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H47F5O3S
CAS Number:
Molecular Weight:
606.77
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

>98% (HPLC)

form

powder

solubility

DMSO: >5 mg/mL

originator

AstraZeneca

storage temp.

2-8°C

SMILES string

[H][C@]12CC[C@]3(C)[C@@H](O)CC[C@@]3([H])[C@]1([H])[C@H](CCCCCCCCCS(=O)CCCC(F)(F)C(F)(F)F)Cc4cc(O)ccc24

InChI

1S/C32H47F5O3S/c1-30-17-15-26-25-12-11-24(38)21-23(25)20-22(29(26)27(30)13-14-28(30)39)10-7-5-3-2-4-6-8-18-41(40)19-9-16-31(33,34)32(35,36)37/h11-12,21-22,26-29,38-39H,2-10,13-20H2,1H3/t22-,26-,27+,28+,29-,30+,41?/m1/s1

InChI key

VWUXBMIQPBEWFH-WCCTWKNTSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Fulvestrant has been used:
  • as an antagonist of estrogen receptor (ERα and ERβ) to reduce hepatocytes proliferation
  • as an antagonist of ER for the suppression of estrogen responsive element in human breast cancer MCF-7 cells
  • as anti-estrogen in breast cancer cell lines

Biochem/physiol Actions

Fulvestrant (ICI 182,780) is a selective estrogen receptor down-regulator (SERD). Fulvestrant is a high affinity estrogen receptor antagonist. IC50 = 0.29 nM. Fulvestrant is the first "pure" antiestrogen with no agonistic activity both in vitro and in vivo.
Fulvestrant is a 7α-alkylsulphinyl analog of 17β-oestradiol and is structurally different compared to other selective estrogen receptor (ER) modulators (SERMs). It reduces dimerization and nuclear localization of the estrogen receptor (ER). Fulvestrant lowers the level of ER protein in human breast cancer cells. Fulvestrant is preferred for the treatment of women at their menopause having hormone-sensitive advanced breast cancer.
Fulvestrant is a selective estrogen receptor down-regulator (SERD); high affinity estrogen receptor antagonist.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Lact. - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glutamic acid ameliorates estrogen deficiency-induced menopausal-like symptoms in ovariectomized mice
Han NR, et al.
Nutrition Research (New York, N.Y.), 35(9), 774-783 (2015)
Fulvestrant: an oestrogen receptor antagonist with a novel mechanism of action
Osborne CK, et al.
British Journal of Cancer, 90(S1), S2-S2 (2004)
Nuclear Respiratory Factor 1 Acting as an Oncoprotein Drives Estrogen-Induced Breast Carcinogenesis
Das J, et al.
Cells, 7(12), 234-234 (2018)
Partial hepatectomy induces delayed hepatocyte proliferation and normal liver regeneration in ovariectomized mice
Umeda M, et al.
Clinical and Experimental Gastroenterology, 8(2), 175-175 (2015)
Manuele G Muraro et al.
Oncoimmunology, 6(7), e1331798-e1331798 (2017-08-16)
Interaction between cancerous, non-transformed cells, and non-cellular components within the tumor microenvironment plays a key role in response to treatment. However, short-term culture or xenotransplantation of cancer specimens in immunodeficient animals results in dramatic modifications of the tumor microenvironment, thus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service