Skip to Content
Merck
All Photos(1)

Key Documents

SML1311

Sigma-Aldrich

Rubone

≥98% (HPLC)

Synonym(s):

(2E)-1-(2-Hydroxy-4,6-dimethoxyphenyl)-3-(2,4,5-trimethoxyphenyl)-2-propen-1-one, 2′-Hydroxy-2,4,4′,5,6′-pentamethoxychalcone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22O7
CAS Number:
Molecular Weight:
374.38
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

yellow to orange

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O(C)c1c(cc(c(c1)OC)\C=C\C(=O)c2c(cc(cc2O)OC)OC)OC

InChI

1S/C20H22O7/c1-23-13-9-15(22)20(19(10-13)27-5)14(21)7-6-12-8-17(25-3)18(26-4)11-16(12)24-2/h6-11,22H,1-5H3/b7-6+

InChI key

VHCQVGQULWFQTM-VOTSOKGWSA-N

Biochem/physiol Actions

Rubone has strong anticancer activity. Rubone and paclitaxel (PTX) combination therapy retarded cancer cell growth, migration and cancer stem-like cells (CSC) population growth.
Rubone is a miR34a modulator that specifically restores miR34a in hepatocellular carcinoma cells with wild-type or mutated p53. Rubone potently inhibits growth of hepatocellular carcinoma in a mouse xenograft model. Rubone does not effect the growth of nontumorigenic human hepatocytes.
Rubone is a miR34a modulator that specifically restores miR34a in hepatocellular carcinoma cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chong Chen et al.
Oncogene, 38(23), 4527-4539 (2019-02-12)
The altered metabolism and acidic microenvironment plays an important role in promoting tumor malignant characteristics. A small population of cancer stem cells (CSCs) were considered as a therapy target to reserve tumor relapse, resistance, and metastasis. However, the molecular mechanism
Micellar delivery of miR-34a modulator rubone and paclitaxel in resistant prostate cancer
Wen D, et al.
Cancer Research, 77(12), 3244-3254 (2017)
Small molecule targeting miR-34a for cancer therapy
Xiao Z, et al.
Molecular & Cellular Oncology, 2(1), e977160-e977160 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service