Skip to Content
Merck
All Photos(1)

Key Documents

SML1144

Sigma-Aldrich

Noopept

≥98% (HPLC)

Synonym(s):

1-(2-Phenylacetyl)-L-prolyl-glycine ethyl ester, GVS 111, N-Phenylacetyl-L-prolylglycine ethyl ester, SGS 111

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22N2O4
CAS Number:
Molecular Weight:
318.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -115 to -125°, c = 0.5 in chloroform-d

color

white to beige

solubility

DMSO: 20 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

O=C(CC1=CC=CC=C1)N2CCC[C@H]2C(NCC(OCC)=O)=O

InChI

1S/C17H22N2O4/c1-2-23-16(21)12-18-17(22)14-9-6-10-19(14)15(20)11-13-7-4-3-5-8-13/h3-5,7-8,14H,2,6,9-12H2,1H3,(H,18,22)/t14-/m0/s1

InChI key

PJNSMUBMSNAEEN-AWEZNQCLSA-N

Biochem/physiol Actions

Noopept is a nootropic and neuroprotective drug that normalizes the balance of the pro- and antioxidant systems.
Noopept is a nootropic and neuroprotective drug that normalizes the balance of the pro- and antioxidant systems. Noopept modulates a variety of physiological functions including cognition and anxiety. Noopept significantly weakens streptozotocin-Induced diabetes in rats.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service