Skip to Content
Merck
All Photos(1)

Key Documents

SML1004

Sigma-Aldrich

ML211

Synonym(s):

4-tert-Butylpiperidin-1-yl-[4-[hydroxy(diphenyl)methyl]triazol-2-yl]methanone, CID-56593118

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H30N4O2
Molecular Weight:
418.53
UNSPSC Code:
41106300
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

ML211 is a very potent, dual inhbitor of lysophospholipase 1 (LYPLA1; IC50 = 17 nM) and lysophospholipase 2 (LYPLA2; IC50 = 30 nM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Cravatt group develops innovative technologies to understand enzyme roles in disease, focusing on activity-based protein profiling.

The Cravatt group develops innovative technologies to understand enzyme roles in disease, focusing on activity-based protein profiling.

The Cravatt group develops innovative technologies to understand enzyme roles in disease, focusing on activity-based protein profiling.

The Cravatt group develops innovative technologies to understand enzyme roles in disease, focusing on activity-based protein profiling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service