Skip to Content
Merck
All Photos(1)

Key Documents

SML0935

Sigma-Aldrich

Aripiprazole

≥98% (HPLC)

Synonym(s):

7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-2(1H)-quinolinone, 7-{4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy}-3,4-dihydro-2(1H)-quinolinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27Cl2N3O2
CAS Number:
Molecular Weight:
448.39
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

Clc1c(cccc1N2CCN(CC2)CCCCOc3cc4c(cc3)CCC(=O)N4)Cl

InChI

1S/C23H27Cl2N3O2/c24-19-4-3-5-21(23(19)25)28-13-11-27(12-14-28)10-1-2-15-30-18-8-6-17-7-9-22(29)26-20(17)16-18/h3-6,8,16H,1-2,7,9-15H2,(H,26,29)

InChI key

CEUORZQYGODEFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3, 4 dihydro-2--(1H)quinolinone is the major component of aripiprazole.[1]

Application

Aripiprazole has been used to study behavioral and neurochemical effects of aripiprazole treatment in juvenile spontaneously hypertensive rats (SHR), a model for attention deficit hyperactivity disorder (ADHD).[2]
Aripiprazole has been used to study its effect on mammary tumorigenesis, mediated by retrovirus delivery of receptor tyrosine-protein kinase 2 or HRas proto-oncogene, GTPase or by transgenic expression of Wnt-1 (proto-oncoprotein).[3] It has also been used to study its behavioral and neurochemical effects in juvenile spontaneously hypertensive rats.[4]

Biochem/physiol Actions

Aripiprazole is a second generation atypical antipsychotic and anti-depressant with partial agonist activity at dopamine D2 and serotonin 5-HT1A receptors and antagonist activity at serotonin 5-HT2A receptors.[5][6] Ki values are 0.34 nM, 0.8 nM, 1.7 nM, and 3.4 nM, respectively, for dopamine D2 and D3, serotonin 5-HT1A and 5-HT2A receptors.[6][1] Aripiprazole is used in the treatment of schizophrenia.[4]
Aripiprazole serves as a ‘dopaminergic stabilizer′. It is used as a potential therapeutic to treat schizophrenia patients. In addition, it is also preferably used to treat attention deficit hyperactivity disorder (ADHD) in patients with Tourette syndrome (TS).[2]

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neil M Richtand et al.
Physiology & behavior, 106(2), 171-177 (2012-02-22)
Studies characterizing treatment interventions in a naturalistic setting suggest that antidepressant and antipsychotic medications may be equally effective in improving clinical outcome in individuals at high risk for first-episode psychosis. Of interest, both beneficial as well as potentially adverse effects
Aripiprazole and Riluzole treatment alters behavior and neurometabolites in young ADHD rats: a longitudinal 1 H-NMR spectroscopy study at 11.7 T.
Rizzo F, et al.
Translational Psychiatry, 7(8), e1189-e1189 null
D M Taylor
International journal of clinical practice, 57(1), 49-54 (2003-02-18)
Atypical antipsychotics generally have a lower propensity for extrapyramidal side-effects (EPSE), hyperprolactinaemia and tardive dyskinesia than that associated with typical antipsychotics but may still produce troublesome side-effects, such as weight gain, cardiac rhythm changes and impaired glucose tolerance. Aripiprazole is
Hyperprolactinemia-inducing antipsychotics increase breast cancer risk by activating JAK-STAT5 in precancerous lesions.
Johnston A N, et al.
Breast Cancer Research, 20(1), 42-42 (2018)
Aripiprazole, a novel antipsychotic agent: dopamine D2 receptor partial agonist.
Hirose T and Kikuchi T
The Journal of Medical Investigation, 52(Supplement), 284-290 (2005)

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service