Skip to Content
Merck
All Photos(1)

Key Documents

SML0527

Sigma-Aldrich

Epalrestat

≥98% (HPLC)

Synonym(s):

(5Z)-5-[(2E)-2-Methyl-3-phenyl-2-propen-1-ylidene]-4-oxo-2-thioxo-3-thiazolidineacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13NO3S2
CAS Number:
Molecular Weight:
319.40
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

yellow to orange

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

S1\C(=C/C(=C/c2ccccc2)/C)\C(=O)N(C1=S)CC(=O)O

InChI

1S/C15H13NO3S2/c1-10(7-11-5-3-2-4-6-11)8-12-14(19)16(9-13(17)18)15(20)21-12/h2-8H,9H2,1H3,(H,17,18)/b10-7+,12-8-

InChI key

CHNUOJQWGUIOLD-NFZZJPOKSA-N

Application

Epalrestat has been used as an aldose reductase inhibitor:
  • in the dahomey larvae diet fed forDrosophila
  • for non-irradiated and X-ray irradiated human aldose reductase
  • to test its protective effect in mice with bleomycin-induced pulmonary fibrosis

Biochem/physiol Actions

Epalrestat inhibits Aldose Reductase (AR) involved in the rate limiting step in the conversion of glucose to sorbitol under hyperglycemic conditions. Aldose reductase has been the target of multiple clinical investigatons to treat diabetic neuropathy and retinopathy. Epalrestat is an approved drug in Japan and India, used for the treatment of diabetic neuropathy.
Epalrestat is an Aldose Reductase inhibitor.

Features and Benefits

This compound is featured on the Dopamine and Norepinephrine Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Other Notes

Light sensitve

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyun Ah Jung et al.
The Journal of pharmacy and pharmacology, 60(9), 1227-1236 (2008-08-23)
Important targets for the prevention and treatment of diabetic complications include aldose reductase (AR) inhibitors (ARIs) and inhibitors of advanced glycation endproduct (AGE) formation. Here we evaluate the inhibitory activities of prenylated flavonoids isolated from Sophora flavescens, a traditional herbal
Juan Gu et al.
Journal of ethnopharmacology, 133(1), 6-13 (2010-09-08)
To investigate the effects of lignans extracted from Eucommia ulmoides and epalrestat on vascular remodeling in spontaneously hypertensive rats. Ten-week-old male spontaneously hypertensive rats were randomly divided into 3 groups (12 rats each group), and treated orally with 100 mg/kg/d
Zhen-Yu Li et al.
The American journal of Chinese medicine, 41(4), 801-815 (2013-07-31)
The lignan extracts from the tree bark of Eucommia ulmoides Oliv., a famous traditional Chinese medicine, have been demonstrated to have inhibitory effects on aldose reductase activity in spontaneously hypertensive rat myocardium. This study was aimed to investigate the hypertensive
Mary Ann Ramirez et al.
Pharmacotherapy, 28(5), 646-655 (2008-05-02)
Diabetic neuropathy is one of the most common long-term complications in patients with diabetes mellitus, with a prevalence of 60-70% in the United States. Treatment options include antidepressants, anticonvulsants, tramadol, and capsaicin. These agents are modestly effective for symptomatic relief
J W Steele et al.
Drugs & aging, 3(6), 532-555 (1993-11-01)
Epalrestat is a carboxylic acid derivative which inhibits aldose reductase, an enzyme of the sorbitol (polyol) pathway. Under hyperglycaemic conditions epalrestat reduces intracellular sorbitol accumulation, which has been implicated in the pathogenesis of late-onset complications of diabetes mellitus. Epalrestat 150

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service